Skip to Content
Merck
All Photos(1)

Key Documents

PHR1542

Supelco

Moxifloxacin Hydrochloride

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Moxifloxacin hydrochloride, 1-Cyclopropyl-6-fluoro-8-methoxy-7-[(4aS,7aS)-octahydropyrrolo[3,4-b]pyridin-6-yl]-4-oxo-1,4-dihydroquinoline-3-carboxylic acid hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H24FN3O4· HCl
CAS Number:
Molecular Weight:
437.89
Beilstein:
8377447
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. Y0000703
traceable to USP 1448606

API family

moxifloxacin

CofA

current certificate can be downloaded

packaging

pkg of 1 g

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

Cl.COc1c(N2C[C@@H]3CCCN[C@@H]3C2)c(F)cc4C(=O)C(=CN(C5CC5)c14)C(O)=O

InChI

1S/C21H24FN3O4.ClH/c1-29-20-17-13(19(26)14(21(27)28)9-25(17)12-4-5-12)7-15(22)18(20)24-8-11-3-2-6-23-16(11)10-24;/h7,9,11-12,16,23H,2-6,8,10H2,1H3,(H,27,28);1H/t11-,16+;/m0./s1

InChI key

IDIIJJHBXUESQI-DFIJPDEKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Moxifloxacin Hydrochloride is an anti-infectious compound that belongs to the class of fluoroquinolone antibiotics. It is a well-known broad-spectrum antibiotic prescription medicine effective against a variety of Gram-positive and Gram-negative bacteria including the anaerobic type. It is an S,S-isomer with two chiral centers. It works by preventing the activity of various crucial enzymes involved in the replication, transcription, recombination, and repair of deoxyribonucleic acid in the bacteria such as DNA gyrase (topoisomerase II), topoisomerase IV. It finds its application in the treatment of various ocular and respiratory tract infections.
Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Application

Moxifloxacin Hydrochloride can be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations and combined dosage forms by spectrophotometric, chromatographic, and electrochemical techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC0258 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enantiomeric purity assay of moxifloxacin hydrochloride by capillary electrophoresis
Cruz LA and Hall R
Journal of Pharmaceutical and Biomedical Analysis, 38(1), 8-13 (2005)
Electrochemical determination of antibacterial drug moxifloxacin hydrochloride using chloranil modified carbon paste electrode
Attia AK and El-Shal MA
Analytical and Bioanalytical Chemistry, 4(3), 213-224 (2012)
Development and validation of UV-spectrophotometric methods for determination of moxifloxacin HCl in bulk and pharmaceutical formulations
Tarkase KN, et al.
Der Pharma Chemica, 4(3), 1180-1185 (2012)
Stability indicating HPLC method for the simultaneous determination of moxifloxacin and prednisolone in pharmaceutical formulations
Razaaq NS, et al.
Chemistry Central Journal, 6(1), 94-94 (2012)
Validated spectrophotometric methods for the estimation of moxifloxacin in bulk and pharmaceutical formulations
Motwani SK, et al.
Spectrochimica Acta. Part A, Molecular and Biomolecular Spectroscopy, 68(2), 250-256 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service