Skip to Content
Merck
All Photos(1)

Key Documents

PHR1270

Supelco

Fumaric Acid

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Fumaric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOCCH=CHCOOH
CAS Number:
Molecular Weight:
116.07
Beilstein:
605763
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. F0600000
traceable to USP 1286708

vapor pressure

1.7 mmHg ( 165 °C)

API family

fumaric acid

CofA

current certificate can be downloaded

autoignition temp.

1364 °F

expl. lim.

40 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

298-300 °C (subl.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OC(=O)\C=C\C(O)=O

InChI

1S/C4H4O4/c5-3(6)1-2-4(7)8/h1-2H,(H,5,6)(H,7,8)/b2-1+

InChI key

VZCYOOQTPOCHFL-OWOJBTEDSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Fumaric Acid may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by spectrophotometry.[1][2]
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC1693 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

521.6 - 525.2 °F - open cup

Flash Point(C)

272 - 274 °C - open cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Spectrophotometric estimation of ketotifen fumarate in pharmaceutical formulations
Sastry CSP and Naidu PY
Microchimica Acta, 127(3-4), 219-223 (1997)
Spectrophotometric determination of ascorbic acid in pharmaceuticals by background correction and flow injection
Verma KK, et al.
Analyst, 116(6), 641-645 (1991)
Fumaric acid: a new feed additive for pigs.
Nutrition reviews, 43(8), 253-254 (1985-08-01)
R Gold et al.
Clinical immunology (Orlando, Fla.), 142(1), 44-48 (2011-03-19)
Fumaric acid was originally therapeutically used in psoriasis. Several lines of evidence have demonstrated immunomodulatory but also neuroprotective effects for FAE. Clinical studies in psoriasis showed a reduction of peripheral CD4+ and CD8+ T-lymphocytes due to the ability of FAE
Qing Xu et al.
Biotechnology advances, 30(6), 1685-1696 (2012-09-04)
The growing concern about the safety of food and dairy additives and the increasing costs of petroleum-based chemicals have rekindled the interest in the fermentation processes for fumaric acid production. The key problems of the industrial production of microbial fumaric

Articles

A new RP-HPLC method is proposed for the determination of organic acids in 100% aqueous mobile phase using an Ascentis® Express AQ-C18 column

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service