Skip to Content
Merck
All Photos(1)

Key Documents

P4200000

Pyrimethamine

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

5-(4-Chlorophenyl)-6-ethyl-2,4-pyrimidinediamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H13ClN4
CAS Number:
Molecular Weight:
248.71
Beilstein:
219864
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

pyrimethamine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CCC1=NC(N)=NC(N)=C1C2=CC=C(Cl)C=C2

InChI

1S/C12H13ClN4/c1-2-9-10(11(14)17-12(15)16-9)7-3-5-8(13)6-4-7/h3-6H,2H2,1H3,(H4,14,15,16,17)

InChI key

WKSAUQYGYAYLPV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Pyrimethamine EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna Maria van Eijk et al.
The Lancet. Infectious diseases, 13(12), 1029-1042 (2013-09-24)
Pregnant women in malaria-endemic countries in sub-Saharan Africa are especially vulnerable to malaria. Recommended prevention strategies include intermittent preventive treatment with two doses of sulfadoxine-pyrimethamine and the use of insecticide-treated nets. However, progress with implementation has been slow and the
Inbarani Naidoo et al.
Trends in parasitology, 29(10), 505-515 (2013-09-14)
Sulfadoxine-pyrimethamine (SP) is used throughout Africa for intermittent preventive treatment (IPT) of malaria, but resistance threatens its efficacy. We found marked regional differences in the genotypes responsible for SP resistance when mapping recent surveys of dihydrofolate reductase (dhfr) and dihydropteroate
Julie Gutman et al.
The Journal of infectious diseases, 208(6), 907-916 (2013-06-27)
Malaria during pregnancy is associated with low birth weight and increased perinatal mortality, especially among primigravidae. Despite increasing prevalence of malarial parasite resistance to sulfadoxine-pyrimethamine (SP), SP continues to be recommended for intermittent preventive treatment in pregnancy (IPTp). Women without
Meera Venkatesan et al.
Trends in parasitology, 29(10), 497-504 (2013-08-21)
Mutations in the Plasmodium falciparum genes Pfdhfr and Pfdhps have rendered sulfadoxine-pyrimethamine (SP) ineffective for malaria treatment in most regions of the world. Yet, SP is efficacious as intermittent preventive therapy in pregnant women (IPTp) and infants (IPTi) and as
Mari Luntamo et al.
Tropical medicine & international health : TM & IH, 18(4), 386-397 (2013-02-26)
To examine the potential to reduce foetal and neonatal growth faltering through intermittent preventive treatment in pregnancy (IPTp) of malaria and reproductive tract infections with monthly sulphadoxine-pyrimethamine (SP), alone or with two doses of azithromycin. We enrolled 1320 women with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service