Skip to Content
Merck
All Photos(1)

Key Documents

P2600000

Poly(vinyl chloride)

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

PVC

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH2CHCl)n
CAS Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

manufacturer/tradename

EDQM

density

1.4 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

ClC=C

InChI

1S/C2H3Cl/c1-2-3/h2H,1H2

InChI key

BZHJMEDXRYGGRV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Poly(vinyl chloride) EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christopher B Stipe et al.
Applied spectroscopy, 66(11), 1286-1293 (2012-11-14)
Airborne silica dust (quartz) is common in coal mines and represents a respiratory hazard that can lead to silicosis, a potentially fatal lung disease. With an eye toward developing a portable monitoring device for rapid analysis of silica dust, laser-induced
T D Ambrisko et al.
British journal of anaesthesia, 110(2), 305-310 (2012-11-21)
In a previous study, the authors found a large bias (50%) for lithium (LiDCO) compared with thermodilution cardiac output measurement methods in ponies receiving i.v. infusions of xylazine, ketamine, and midazolam. This prompted the authors to examine the effect of
Olivier Firmesse et al.
International journal of food microbiology, 159(3), 179-185 (2012-10-31)
The aim of this study was to determine how quickly the surface of a refrigerated supermarket serve over counter becomes loaded with bacteria. New material made of polyvinyl chloride or stainless steel was placed on the surface on which foodstuffs
Gianluigi Li Bassi et al.
Critical care medicine, 41(2), 518-526 (2012-12-25)
To assess the structural characteristics involved in the design of high-volume low-pressure endotracheal tube cuffs that are associated with fluid sealing effectiveness and to determine the extent of transmitted tracheal pressures upon cuff inflation. In vitro study. Pneumology laboratories. Eight
Yujie Lei et al.
Zhongguo xiu fu chong jian wai ke za zhi = Zhongguo xiufu chongjian waike zazhi = Chinese journal of reparative and reconstructive surgery, 26(9), 1117-1121 (2012-10-13)
To explore the influence of three central venous catheter biomedical materials (polyurethane, silicone, and polyvinyl chloride) on the proliferation, apoptosis, and cell cycle of Xuanwei Lung Cancer-05 (XWLC-05) cells so as to provide the basis for clinical choice of central

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service