Skip to Content
Merck
All Photos(1)

Key Documents

P1000000

Phenoxymethylpenicillin

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Phenoxymethylpenicillinic acid, Penicillin V

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H18N2O5S
CAS Number:
Molecular Weight:
350.39
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

penicillin & derivatives

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC1(C)S[C@@H]2[C@H](NC(=O)COc3ccccc3)C(=O)N2[C@H]1C(O)=O

InChI

1S/C16H18N2O5S/c1-16(2)12(15(21)22)18-13(20)11(14(18)24-16)17-10(19)8-23-9-6-4-3-5-7-9/h3-7,11-12,14H,8H2,1-2H3,(H,17,19)(H,21,22)/t11-,12+,14-/m1/s1

InChI key

BPLBGHOLXOTWMN-MBNYWOFBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Phenoxymethylpenicillin EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rakesh Gopal et al.
Indian heart journal, 64(1), 12-15 (2012-05-11)
Rheumatic fever and rheumatic heart disease (RHD) are still important problems in developing countries. Secondary prophylaxis which is the most cost-effective method in preventing recurrences of rheumatic fever is fraught with problems of drug compliance. The utility of 500 mg
Editors' comments.
Joel Schwab et al.
Acta paediatrica (Oslo, Norway : 1992), 101(7), 705-705 (2012-06-08)
Manish Soneja et al.
The Journal of the Association of Physicians of India, 60, 66-68 (2012-10-04)
Chronic granulomatous disease (CGD) is an inherited disease of the phagocyte NADPH oxidase system that causes defective production of toxic oxygen metabolites, leading to impaired bacterial and fungal killing, and recurrent life threatening infections; mostly by catalase producing organisms. Nocardiosis
Michael Quirke et al.
Trials, 14, 164-164 (2013-06-05)
Oral flucloxacillin, either alone or in combination with phenoxymethylpenicillin, is a commonly prescribed antibiotic for the treatment of cellulitis, particularly in Ireland and the United Kingdom. This study aims to establish the non-inferiority of oral monotherapy (flucloxacillin alone) to dual
Cristina Giovannoli et al.
Journal of molecular recognition : JMR, 25(6), 377-382 (2012-05-30)
Molecularly imprinted polymers have been successfully used as selective stationary phases in capillary electrophoresis. Notwithstanding, this technique suffers from several drawbacks as the loss of molecular recognition properties in aqueous media and the lack of feasibility for imprinted systems directed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service