Skip to Content
Merck
All Photos(2)

Key Documents

NIST937

Iron metal

NIST® SRM® 937

Synonym(s):

Iron

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Fe
CAS Number:
Molecular Weight:
55.85
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

solid

shelf life

10 yr

packaging

pkg of 50 g

manufacturer/tradename

NIST®

resistivity

9.71 μΩ-cm

bp

2750 °C (lit.)

mp

1535 °C (lit.)

density

7.86 g/mL at 25 °C (lit.)

application(s)

clinical

format

matrix material

SMILES string

[Fe]

InChI

1S/Fe

InChI key

XEEYBQQBJWHFJM-UHFFFAOYSA-N

General description

This certified standard reference material (SRM) consists of electrolytic grade iron and is a material of known purity. A unit of NIST SRM 937 consists of 50 g of iron metal in the form of chips sized between 0.5 mm and 1.8 mm mesh. For more information, please refer to the COA and SDS.

SRM 937_cert


SRM 937 _SDS

Application

This SRM 937 is intended for use as an assay standard for iron. It is primarily used for the calibration of instrumentation and standardization of procedures and for the routine evaluation of daily working standards used in these procedures.

Other Notes

  • This SRM should be stored in the tightly closed original bottle under normal laboratory conditions.
  • Expiration details and storage and use instructions are provided on the NIST certificate
  • Notes on biomaterial handling, disposal, and transport documented in MSDS
  • This SRM is intended for research use.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Iron and zinc levels in Vigna unguiculata (L.) Walp under varying phosphorus and fixed nitrogen treatment conditions.
Ayeni F D, et al.
Food and energy security, e00141-e00141 (2018)
Katsuyuki Shinohara et al.
Clinical chemistry, 62(2), 392-406 (2015-12-17)
The Japanese Committee for Clinical Laboratory Standards (JCCLS) has developed a multianalyte conventional reference material (MacRM) for nationwide standardization of laboratory measurements. To prepare the MacRM, pooled sera were obtained from healthy Japanese individuals. Target values of the pooled sera
Architectural Graphic Standards for Residential Construction (2010)
Renata Paleari et al.
Clinica chimica acta; international journal of clinical chemistry, 541, 117246-117246 (2023-02-09)
Glycated albumin (GA) may assess glycometabolic control over a short period of time respect to HbA1c, and its use to screen for gestational diabetes in pregnancy has been suggested. To this regard few data on reference intervals (RI) for GA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service