Skip to Content
Merck
All Photos(1)

Documents

NIST1595

Tripalmitin

NIST® SRM® 1595

Synonym(s):

Tripalmitin, 1,2,3-Propanetriol tris(hexadecanoate), Glyceryl tripalmitate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)14COOCH2]2CHOCO(CH2)14CH3
CAS Number:
Molecular Weight:
807.32
Beilstein:
1811188
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 2 g

manufacturer/tradename

NIST®

mp

66-67 °C (lit.)

format

neat

SMILES string

CCCCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCCCCC)OC(=O)CCCCCCCCCCCCCCC

InChI

1S/C51H98O6/c1-4-7-10-13-16-19-22-25-28-31-34-37-40-43-49(52)55-46-48(57-51(54)45-42-39-36-33-30-27-24-21-18-15-12-9-6-3)47-56-50(53)44-41-38-35-32-29-26-23-20-17-14-11-8-5-2/h48H,4-47H2,1-3H3

InChI key

PVNIQBQSYATKKL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Other Notes

Certified for the analytes listed below.
Tripalmitin

Reference/Informational value is provided for the following analytes.
Unknown glyceride, Methanol (CH3OH), Insoluble matter, Residue on ignition, Carbon (C), Hydrogen (H)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abd El-Halim I El Assasy et al.
AAPS PharmSciTech, 20(2), 82-82 (2019-01-18)
Amisulpride (AMS), a second generation antipsychotic, suffers from low oral bioavailability (48%). This might be due to its pH-dependent solubility or being a substrate of P-glycoprotein efflux pump. Nanostructured lipid carriers (NLCs) were proposed in this study to enhance the
Aaron Millan-Oropeza et al.
Applied spectroscopy, 71(10), 2344-2352 (2017-05-10)
There is a growing interest worldwide for the production of renewable oil without mobilizing agriculture lands; fast and reliable methods are needed to identify highly oleaginous microorganisms of potential industrial interest. The aim of this study was to demonstrate the
Isabelle J Gabriëls et al.
International journal of molecular sciences, 20(6) (2019-03-27)
In Europe, the toxicological safety of genetically modified (GM) crops is routinely evaluated using rodent feeding trials, originally designed for testing oral toxicity of chemical compounds. We aimed to develop and optimize methods for advancing the use of zebrafish feeding
Yurika Otoki et al.
Lipids, 52(6), 559-571 (2017-04-26)
Disturbances in peripheral and brain lipid metabolism, including the omega-3 fatty acid docosahexaenoic acid (DHA), have been reported in major depressive disorder (MDD). However, these changes have yet to be confirmed in MDD with seasonal pattern (MDD-s), a subtype of
Takashi Yada et al.
Comparative biochemistry and physiology. Part A, Molecular & integrative physiology, 245, 110700-110700 (2020-04-16)
To clarify the relationships between growth, endocrine status and habitat characteristics in Japanese eel (Anguilla japonica), plasma and stomach mRNA levels of ghrelin were examined in wild eels captured in the river and the bay, and in cultured eels during

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service