Skip to Content
Merck
All Photos(1)

Key Documents

NIST1224

Low alloy steel

NIST® SRM® 1224, carbon (AISI 1078)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of disk

manufacturer/tradename

NIST®

format

matrix material

Looking for similar products? Visit Product Comparison Guide

General description

A unit of SRM 1224 comprises a single disk measuring 32 mm in diameter and 19 mm in thickness. For more information, please refer to the SDS and COA.
SRM 1224_cert
SRM 1224_SDS

Application

Low alloy steel Standard Reference Material (SRM) is primarily intended for validating chemical and instrumental methods of analysis.

Features and Benefits

  • Certified and reference mass fraction values for various constituents are provided, values are metrologically traceable to the SI unit of mass and estimated using a consistent statistical approach to determine measurement uncertainty.
  • SRM 1224 provides certified mass fraction values, indicating the highest level of accuracy.
  • Non-certified values are presented as the best estimates of true values for the reference mass fraction, along with their associated uncertainties.
  • Expiration details and instructions for use are provided on the NIST certificate.

Other Notes

Certified for the analytes listed below.
Carbon (C), Chromium (Cr), Copper (Cu), Manganese (Mn), Molybdenum (Mo), Nickel (Ni), Phosphorus (P), Silicon (Si), Sulfur (S)

Reference/Informational value is provided for the following analytes.
Aluminum (Al), Vanadium (V)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service