Skip to Content
Merck
All Photos(3)

Documents

M7554

Sigma-Aldrich

Magnesium D-gluconate hydrate

98.0-102% anhydrous basis, meets USP testing specifications

Synonym(s):

D-Gluconic acid hemimagnesium salt, D-Gluconic acid magnesium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22MgO14 · xH2O
CAS Number:
Molecular Weight:
414.60 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic (organic)

Quality Level

Agency

USP/NF
meets USP testing specifications

Assay

98.0-102% anhydrous basis

form

solid

impurities

3.0-12.0% water (Karl Fischer)

color

white

useful pH range

6.0 - 7.8

mp

169.67 - 171.93  °C ((337.41 - 341.47 °F ))

solubility

9.6 g/L at 20 °C

application(s)

pharmaceutical (small molecule)

storage temp.

room temp

SMILES string

O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C(=O)O[Mg]OC(=O)[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO

InChI

1S/2C6H12O7.Mg.H2O/c2*7-1-2(8)3(9)4(10)5(11)6(12)13;;/h2*2-5,7-11H,1H2,(H,12,13);;1H2/q;;+2;/p-2/t2*2-,3-,4+,5-;;/m11../s1

InChI key

IAKLPCRFBAZVRW-XRDLMGPZSA-L

Looking for similar products? Visit Product Comparison Guide

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abdelali Daddaoua et al.
PloS one, 7(7), e39390-e39390 (2012-07-31)
Homologs of the transcriptional regulator PtxS are omnipresent in Pseudomonas, whereas PtxR homologues are exclusively found in human pathogenic Pseudomonas species. In all Pseudomonas sp., PtxS with 2-ketogluconate is the regulator of the gluconate degradation pathway and controls expression from
Angel A Yanagihara et al.
PloS one, 7(12), e51368-e51368 (2012-12-20)
Chironex fleckeri (Australian box jellyfish) stings can cause acute cardiovascular collapse and death. We developed methods to recover venom with high specific activity, and evaluated the effects of both total venom and constituent porins at doses equivalent to lethal envenomation.
Maria Meyer et al.
Applied microbiology and biotechnology, 97(8), 3457-3466 (2012-07-14)
The acetic acid bacterium Gluconobacter oxydans incompletely oxidizes carbon sources as a natural part of its metabolism, and this feature has been exploited for many biotechnological applications. The most important enzymes used to harness the biocatalytic oxidative capacity of G.
Anja Troge et al.
International journal of medical microbiology : IJMM, 302(7-8), 304-314 (2012-11-08)
The flagellum of the probiotic Escherichia coli strain Nissle 1917 (EcN) is not just responsible for motility, but also for EcN's ability to induce the production of human β-defensin 2. Here, we report a third function of this EcN organell.
Ze-Ping Hu et al.
Journal of proteome research, 11(12), 5903-5913 (2012-10-20)
Identification of molecular mechanisms underlying early stage Alzheimer's disease (AD) is important for the development of new therapies against and diagnosis of AD. In this study, nontargeted metabonomics of TASTPM transgenic AD mice was performed. The metabolic profiles of both

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service