Skip to Content
Merck
All Photos(1)

Key Documents

M0370000

Mepivacaine hydrochloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

1-Methyl-2′,6′-pipecoloxylidine hydrochloride, N-(2,6-Dimethylphenyl)-1-methyl-2-piperidinecarboxamide hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H22N2O · HCl
CAS Number:
Molecular Weight:
282.81
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

mepivacaine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CN1CCCCC1C(NC2=C(C)C=CC=C2C)=O.Cl

InChI

1S/C15H22N2O.ClH/c1-11-7-6-8-12(2)14(11)16-15(18)13-9-4-5-10-17(13)3;/h6-8,13H,4-5,9-10H2,1-3H3,(H,16,18);1H

InChI key

RETIMRUQNCDCQB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Mepivacaine hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Local anesthetic. Reversibly blocks transient Na+ inward current, as well as the steady-state K+ outward current.
Local anesthetic. Reversibly blocks transient Na+ inward current, as well as the steady-state K+ outward current. Blocks tandem pore (TASK) and Kv1.5, potassium channels in model systems.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Marhofer et al.
British journal of anaesthesia, 113(1), 177-185 (2014-02-28)
The relation between the pattern of local anaesthetic (LA) spread and the quality of peripheral nerve block is unclear. Twenty-one volunteers were randomized to receive a median nerve block with intended circumferential or intended non-circumferential spread of LA. Different predetermined
Local anesthetic systemic toxicity after combined psoas compartment-sciatic nerve block: analysis of decision factors and diagnostic delay.
Marissa G Vadi et al.
Anesthesiology, 120(4), 987-996 (2014-04-04)
C Aumeier et al.
British journal of anaesthesia, 112(4), 735-741 (2013-10-31)
The use of lipid emulsions to reduce cardiac toxicity of local anaesthetics (LAs) has shown success in experimental studies and some clinical cases, and thus has been implemented in clinical practice. However, lipid treatment is usually given after the occurrence
Gianluca Cappelleri et al.
Anesthesia and analgesia, 119(2), 489-493 (2014-06-11)
Among the various factors influencing the success rate, onset time, and duration of peripheral nerve blocks, the role of local anesthetics concentration remains uncertain. In this prospective, randomized, single-blinded study, we evaluated whether varying the dilution of a fixed dose
Hiroto Yamamoto et al.
Anesthesia and analgesia, 119(6), 1442-1448 (2014-10-01)
It is believed that local anesthetic injected to obtain circumferential spread around nerves produces a more rapid onset and successful blockade after some ultrasound-guided peripheral nerve blocks. However, evidence demonstrating this point is limited only to the popliteal sciatic nerve

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service