Skip to Content
Merck
All Photos(1)

Key Documents

M0240000

Medronic acid

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Methylenediphosphonic acid, MDP, Medronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2[P(O)(OH)2]2
CAS Number:
Molecular Weight:
176.00
Beilstein:
1708494
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

medronic acid

manufacturer/tradename

EDQM

mp

197-199 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

OP(O)(=O)CP(O)(O)=O

InChI

1S/CH6O6P2/c2-8(3,4)1-9(5,6)7/h1H2,(H2,2,3,4)(H2,5,6,7)

InChI key

MBKDYNNUVRNNRF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Medronic acid EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Claude Grison et al.
Carbohydrate research, 341(9), 1117-1129 (2006-04-19)
A direct and general access to D-glycosyl 3-, 5-, or 6-methylenediphosphonates, di-D-glycosyl 1,5-, 3,5-, 3,6-, 5,5-, or 6,6-methylenediphosphonates and dithymidine 3',5'-methylenediphosphonate is described. The method involves the one-pot alkylidenediphosphorylation of glycosyl or thymidine derivatives. No antiviral activity was detected against
E F Petersen et al.
Magnetic resonance in medicine, 44(3), 367-372 (2000-09-07)
31P NMR was used to measure the concentrations and spin-lattice relaxation times of phosphorus-containing metabolites in neocartilage developing in an NMR-compatible hollow-fiber bioreactor over four weeks. Separate studies were performed for tissue developing from chondrocytes taken from the proximal and
Ewa Matczak-Jon et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 11(8), 2357-2372 (2005-01-26)
The crystal structures of 3-methylpiperid-1-ylmethane-1,1-diphosphonic (2), 4-methylpiperid-1-ylmethane-1,1-diphosphonic (3), 2-ethylpiperid-1-ylmethane-1,1-diphosphonic (4), and 2-methylpiperid-1-ylmethane-1,1-diphosphonic (5) acids have been determined and are discussed with respect to their molecular organization and crystal-packing preferences. The chair conformation, predominant also in solution, favors equatorial positioning of
Hao Shen et al.
Journal of clinical microbiology, 53(3), 777-781 (2014-12-19)
Prosthetic joint infection (PJI) is a rare but refractory complication of arthroplasty. Accurate identification of pathogens is a key step for successful treatment of PJI, which remains a challenge for clinicians and laboratory workers. We designed a combined culture method
Daocheng Wu et al.
Biomedical microdevices, 14(3), 497-510 (2012-01-27)
Methylenediphosphonate (MDP)-conjugated adriamycin liposomes (MDP-LADMs) were prepared using mild dynamic dialysis equilibrium method, and their targeted therapeutic effects against osteosarcomas and metastatic SOSP-M lung nodules were evaluated in vivo. The drug loading and encapsulation efficiency of the MDP-LADMs were measured

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service