Skip to Content
Merck
All Photos(1)

Key Documents

M0120010

Malathion impurity B

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Malaoxon, Diethyl 2-[(dimethoxyphosphoryl)sulfanyl]succinate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H19O7PS
CAS Number:
Molecular Weight:
314.29
Beilstein:
1804523
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

malathion

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CCOC(=O)CC(SP(=O)(OC)OC)C(=O)OCC

InChI

1S/C10H19O7PS/c1-5-16-9(11)7-8(10(12)17-6-2)19-18(13,14-3)15-4/h8H,5-7H2,1-4H3

InChI key

WSORODGWGUUOBO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Malathion impurity B EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 3 Oral - Aquatic Chronic 2 - Carc. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

>212.0 °F - closed cup

Flash Point(C)

> 100 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Padilla et al.
Journal of toxicology and environmental health. Part A, 67(18), 1477-1489 (2004-09-17)
Some, but not all, organophosphorus pesticides are more acutely toxic to the young as compared to adults. We have developed an in vitro assay that measures the detoxification potential (via carboxylesterase and A-esterases) of tissues. Previous results using this in
Pergentino Balbuena et al.
Toxicological sciences : an official journal of the Society of Toxicology, 114(2), 260-271 (2010-01-13)
Toxicity and integrity disruption in response to transport through the blood-brain barrier (BBB) of the organophosphates malathion and malaoxon and heavy metal lead acetate were assessed in two in vitro barrier systems. One system was constructed using bovine brain microvascular
A N Ivanov et al.
Talanta, 85(1), 216-221 (2011-06-08)
A simple and reliable technique has been developed for the construction of an amperometric acetylcholinesterase biosensor based on screen-printed carbon electrodes. For the first time, one-step modification using single-walled carbon nanotubes and Co phtalocyanine has been proposed to decrease the
Olajire A Gbaye et al.
Pest management science, 68(9), 1265-1271 (2012-05-19)
Bruchid beetles, Callosobruchus species, are serious pests of economically important grain legumes; their activity in stores is often controlled by the use of synthetic insecticides. Esterases are known to be involved in insecticide resistance in insects. However, there is a
Agustín Basso et al.
Environmental science and pollution research international, 19(1), 214-225 (2011-07-01)
In this study, we determined normal serum butyrylcholinesterase (BChE) and carboxylesterase (CbE) activities in Tupinambis merianae in order to obtain reference values for organophosphorus pesticide monitoring. Forty-two T. merianae individuals were grouped by sex and size to identify potential differences

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service