Skip to Content
Merck
All Photos(1)

Key Documents

K3752

Supelco

α-Ketoglutaric acid disodium salt hydrate

≥95%

Synonym(s):

2-Oxopentanedioic acid disodium salt hydrate, Sodium 2-oxoglutarate dibasic hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C5H4O5Na2· xH2O
CAS Number:
Molecular Weight:
190.06 (anhydrous basis)
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Assay

≥95%

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

O.O.[Na+].[Na+].[O-]C(=O)CCC(=O)C([O-])=O

InChI

1S/C5H6O5.2Na.2H2O/c6-3(5(9)10)1-2-4(7)8;;;;/h1-2H2,(H,7,8)(H,9,10);;;2*1H2/q;2*+1;;/p-2

InChI key

HSSWOFWCOAQLHZ-UHFFFAOYSA-L

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N A Smirnova et al.
Biochemistry. Biokhimiia, 77(10), 1108-1119 (2012-11-20)
This review describes the catalytic mechanism, substrate specificity, and structural peculiarities of alpha-ketoglutarate dependent nonheme iron dioxygenases catalyzing prolyl hydroxylation of hypoxia-inducible factor (HIF). Distinct localization and regulation of three isoforms of HIF prolyl hydroxylases suggest their different roles in
Piotr Dobrowolski et al.
Nutrition (Burbank, Los Angeles County, Calif.), 29(3), 556-561 (2012-12-12)
Proton-pump inhibitors, such as omeprazole, are widely used in the prevention and treatment of gastroesophageal diseases. However, an association between proton-pump inhibitors and the increased risk of bone fractures has been observed, especially in patients treated for extended periods. Conversely
Michael E Jung et al.
The Journal of organic chemistry, 77(23), 11002-11005 (2012-11-21)
Oxidative cleavage of cycloalkene-1-carboxylates, made from the corresponding carboxylic acids, and subsequent oxidation of the resulting ketoaldehyde afforded the important 1-monoesters of 2-ketoalkanedioic acids. Thus ozonolysis of octyl cyclobutene-1-carboxylate followed by sodium chlorite oxidation afforded the 1-monooctyl 2-ketoglutarate. This is
Kenji Manabe et al.
Microbial cell factories, 12, 18-18 (2013-02-20)
The Bacillus subtilis genome-reduced strain MGB874 exhibits enhanced production of exogenous extracellular enzymes under batch fermentation conditions. We predicted that deletion of the gene for RocG, a bi-functional protein that acts as a glutamate dehydrogenase and an indirect repressor of
Vivian R Moure et al.
Journal of bacteriology, 195(2), 279-286 (2012-11-13)
Fe protein (dinitrogenase reductase) activity is reversibly inactivated by dinitrogenase reductase ADP-ribosyltransferase (DraT) in response to an increase in the ammonium concentration or a decrease in cellular energy in Azospirillum brasilense, Rhodospirillum rubrum, and Rhodobacter capsulatus. The ADP-ribosyl is removed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service