Skip to Content
Merck
All Photos(1)

Key Documents

IRMM526A

Niobium

IRMM®, certified reference material, 0.02 mm foil

Synonym(s):

Columbium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Nb
CAS Number:
Molecular Weight:
92.91
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Agency

IRMM®

manufacturer/tradename

JRC

resistivity

13-16 μΩ-cm, 20°C

bp

4742 °C (lit.)

mp

2468 °C (lit.)

density

8.57 g/mL at 25 °C (lit.)

format

matrix material

SMILES string

[Nb]

InChI

1S/Nb

InChI key

GUCVJGMIXFAOAE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

For more information please see:
IRMM526A

Legal Information

IRMM is a registered trademark of European Commission

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M I Tsindlekht et al.
Journal of physics. Condensed matter : an Institute of Physics journal, 25(8), 085701-085701 (2013-01-31)
We report results of measurements of the ac conductivity of a Nb superconducting thin film in a swept dc magnetic field. In the mixed state the swept dc field creates vortices at the film surface which pass through the film
Christos D Malliakas et al.
Journal of the American Chemical Society, 135(5), 1719-1722 (2013-01-23)
2H-NbSe(2) is a canonical Charge-Density-Wave (CDW) layered material the structural details of which remained elusive. We report the detailed structure of 2H-NbSe(2) below the CDW transition using a (3 + 2)-dimensional crystallographic approach on single crystal X-ray diffraction data collected
Evgeniy Papulovskiy et al.
Physical chemistry chemical physics : PCCP, 15(14), 5115-5131 (2013-03-02)
Ab initio DFT calculations of (93)Nb NMR parameters using the NMR-CASTEP code were performed for a series of over fifty individual niobates, and a good agreement has been found with experimental NMR parameters. New experimental and calculated (93)Nb NMR data
Yoon-Jun Kim et al.
ACS nano, 7(1), 732-739 (2012-12-25)
Imaging the three-dimensional atomic-scale structure of complex interfaces has been the goal of many recent studies, due to its importance to technologically relevant areas. Combining atom-probe tomography and aberration-corrected scanning transmission electron microscopy (STEM), we present an atomic-scale study of
Yan Sheng et al.
Optics express, 21(4), 4475-4480 (2013-03-14)
We propose and fabricate a LiNbO₃-based nonlinear photonic crystal with locally ordered ferroelectric domains. The nonlinearity modulation provides sets of uniformly distributed reciprocal lattice vectors, ensuring broadband high frequency conversion efficiency. Frequency tripling via cascading is demonstrated in the range

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service