Skip to Content
Merck
All Photos(1)

Documents

I4386

Sigma-Aldrich

Iodoacetic acid

≥98.0% (T)

Synonym(s):

2-Iodoacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ICH2CO2H
CAS Number:
Molecular Weight:
185.95
Beilstein:
1739079
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic

Quality Level

Assay

≥98.0% (T)

form

powder or flakes

color

white to yellow

mp

77-79 °C (lit.)

solubility

H2O: soluble, clear to hazy

storage temp.

−20°C

SMILES string

OC(=O)CI

InChI

1S/C2H3IO2/c3-1-2(4)5/h1H2,(H,4,5)

InChI key

JDNTWHVOXJZDSN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Iodoacetic acid has been used as an alkylating agent for antibody AR37, serum proteins and strawberry proteome samples extracted from different stages of ripened fruit.
Reagent for the modification of cysteine residues in proteins.

Biochem/physiol Actions

Iodoacetic acid (IAA) blocks the thiol group of cysteine. IAA inhibits glyceraldehyde-3-phosphate dehydrogenase (G3PDH) by interacting with sulfhydryl group of the active site cysteine. IAA inhibits the progression of solid Ehrlich carcinoma. IAA is one of the iodinated disinfection byproducts in drinking water. It is cytotoxic to mammalian cells.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Drinking water disinfection byproduct iodoacetic acid induces tumorigenic transformation of NIH3T3 cells
Wei X, et al.
Environmental Science & Technology, 47(11), 5913-5920 (2013)
Quantitative proteomic investigation employing stable isotope labeling by peptide dimethylation on proteins of strawberry fruit at different ripening stages
Li L, et al.
Journal of Proteomics, 94(5), 219-239 (2013)
Antitumor activities of iodoacetate and dimethylsulphoxide against solid Ehrlich carcinoma growth in mice
Fahim FA, et al.
Biological Research, 36(2), 253-262 (2003)
Carboxymethylation of cysteine using iodoacetamide/iodoacetic acid
The Protein Protocols Handbook, 455-456 (2002)
Database-independent Protein Sequencing (DiPS) Enables Full-length de Novo Protein and Antibody Sequence Determination
Savidor A, et al.
Molecular and Cellular Proteomics, 16(6), 1151-1161 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service