ERMBF414D
Maize GMO Standard ERM-BF414
ERM®, certified reference material, 1.0% GA21, for the detection of GMO maize (GA21) with PCR
Synonym(s):
GMO Standard ERM-BF414, Maize GA21
About This Item
Recommended Products
grade
certified reference material
Agency
ERM®
quality
for the detection of GMO maize (GA21) with PCR
composition
genetically modified GA21 maize, 1.0%
manufacturer/tradename
JRC
application(s)
cleaning products
cosmetics
food and beverages
personal care
pharmaceutical (small molecule)
format
matrix material
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
1 of 4
This Item | ERMBF414B | ERMBF414C | ERMBF414E |
---|---|---|---|
matrix material | matrix material | matrix material | matrix material |
certified reference material | certified reference material | certified reference material | certified reference material |
2-8°C | 2-8°C | 2-8°C | 2-8°C |
JRC | JRC | JRC | JRC |
ERM® | ERM® | ERM® | ERM® |
Analysis Note
ERMBF414D
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service