Skip to Content
Merck
All Photos(1)

Documents

D3635

Supelco

Lithium 3,5-diiodosalicylate

analytical standard

Synonym(s):

2-Hydroxy-3,5-diiodobenzoic acid lithium salt, 3,5-Diiodo-2-hydroxybenzoic acid lithium salt, 3,5-Diiodosalicylic acid lithium salt, LIS

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H3I2LiO3
CAS Number:
Molecular Weight:
395.85
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

description

anionic

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

storage temp.

−20°C

SMILES string

[Li+].Oc1c(I)cc(I)cc1C([O-])=O

InChI

1S/C7H4I2O3.Li/c8-3-1-4(7(11)12)6(10)5(9)2-3;/h1-2,10H,(H,11,12);/q;+1/p-1

InChI key

HLBRJWWTLIAOTE-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Françoise Imbert-Bismut et al.
Clinica chimica acta; international journal of clinical chemistry, 510, 522-530 (2020-08-01)
Hyperammonemia is neurotoxic and as such can be a medical emergency. Preanalytical factors greatly influence the blood ammonia concentration results. Ammonia concentrations measured in the real life setting of a large hospital after pneumatic transport of blood samples and various
S D Upadhaya et al.
Poultry science, 98(10), 4722-4728 (2019-05-09)
The aim of the present study was to evaluate the efficacy of Bacillus-based probiotic in pullet to lay period. A total of 12-wk-old 384 Hy-line Brown pullets (initial BW of 1.05 kg, 8 replications; 16 birds per replication pen) were used
Guanghua Lu et al.
Scientific reports, 6, 38581-38581 (2016-12-08)
Viruses may induce changes in plant hosts and vectors to enhance their transmission. The white-backed planthopper (WBPH) and brown planthopper (BPH) are vectors of Southern rice black-streaked dwarf virus (SRBSDV) and Rice ragged stunt virus (RRSV), respectively, which cause serious
Ivana Lapić et al.
Biochemia medica, 29(3), 030705-030705 (2019-10-19)
Inappropriate laboratory retesting can be addressed by implementing minimum retesting intervals (MRI). The aim of our study was to assess the effectiveness of the implemented MRI protocol for inpatients. Minimum retesting intervals were applied for 53 laboratory tests. The overall
Bruce Stuart et al.
Health services research, 52(3), 1185-1206 (2016-06-22)
To determine the magnitude and mechanisms of response to Medicare Part D cost sharing by low-income subsidy (LIS) recipients using oral hypoglycemic agents (OHAs) and statins. Medicare data for a 5 percent random sample of beneficiaries with diabetes enrolled in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service