Skip to Content
Merck
All Photos(1)

Key Documents

C8845

Millipore

Casein hydrolysate broth

suitable for microbiology, NutriSelect® Plus

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171607
NACRES:
NA.85
Pricing and availability is not currently available.

form

powder

Quality Level

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

final pH

7.3±0.2 (25 °C)

application(s)

clinical research
food and beverages
life science and biopharma

microbiology

suitability

nonselective for Enterococcus spp.
nonselective for Staphylococcus spp.

Application

Used for production of stapholococcal enterotoxin.

Components

Components g/L:
Casein acid hydrolysate, 20.00
Ferric citrate, 0.025
Potassium phosphate, 2.00
Magnesium sulfate, 0.20
L-Cystine, 0.025
Sodium acetate, 7.00
L-Tryptophan, 0.075
Calcium pantothenate, 0.0005
Thiamine, 0.00004
Nicotinic acid, 0.0012

Preparation Note

Suspend 29.33 g in 1000 ml of distilled water. Mix thoroughly. Heat with frequent agitation and boil for one minute. Dispense as desired. Sterilize by autoclaving at 15 lbs pressure (121°C) for 15 minutes.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service