Skip to Content
Merck
All Photos(1)

Key Documents

C1050000

Chenodeoxycholic acid

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

3α,7α-Dihydroxy-5β-cholanic acid, 5β-Cholanic acid-3α,7α-diol, Chenodiol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H40O4
CAS Number:
Molecular Weight:
392.57
Beilstein:
3219887
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

ursodiol

manufacturer/tradename

EDQM

mp

165-167 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

C[C@H](CCC(O)=O)[C@H]1CC[C@H]2[C@@H]3[C@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3CC[C@]12C

InChI

1S/C24H40O4/c1-14(4-7-21(27)28)17-5-6-18-22-19(9-11-24(17,18)3)23(2)10-8-16(25)12-15(23)13-20(22)26/h14-20,22,25-26H,4-13H2,1-3H3,(H,27,28)/t14-,15+,16-,17-,18+,19+,20-,22+,23+,24-/m1/s1

InChI key

RUDATBOHQWOJDD-BSWAIDMHSA-N

Gene Information

human ... NR1H4(9971)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Chenodeoxycholic acid EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Stark et al.
European journal of biochemistry, 266(1), 209-214 (1999-10-29)
Polypeptides were isolated from human bile by extraction with chloroform/methanol, followed by reversed-phase chromatography in methanol/ethylene chloride and gel filtration in chloroform/methanol. Peptides were characterized by SDS/PAGE, sequence analysis and matrix-assisted laser desorption ionization/time-of-flight mass spectrometry. This identified haemoglobin alpha
Francesco Saverio Di Leva et al.
Journal of medicinal chemistry, 56(11), 4701-4717 (2013-05-10)
Here, we report suvanine, a marine sponge sesterterpene, as an antagonist of the mammalian bile acid sensor farnesoid-X-receptor (FXR). Using suvanine as a template, we shed light on the molecular bases of FXR antagonism, identifying the essential conformational changes responsible
Huang Huang et al.
Journal of medicinal chemistry, 55(16), 7037-7053 (2012-08-07)
LBVS of 12480 in-house compounds, followed by HTRF assay, resulted in one nonsteroidal compound (11) with antagonistic activity against FXR (69.01 ± 11.75 μM). On the basis of 11, 26 new derivatives (12a-z) were designed and synthesized accordingly. Five derivatives
Luciano Adorini et al.
Drug discovery today, 17(17-18), 988-997 (2012-06-02)
Nonalcoholic fatty liver disease (NAFLD) is a highly prevalent chronic liver condition evolving in a proportion of patients into nonalcoholic steatohepatitis (NASH), an aggressive form of NAFLD associated with increased cardiovascular mortality and significant risk of progressive liver disease, including
G S Tint et al.
Gastroenterology, 91(4), 1007-1018 (1986-10-01)
Orally administered UDCA dramatically reduces the secretion of cholesterol into the bile. During UDCA therapy cholesterol balance is maintained by a reduction in both the relative and absolute absorption of cholesterol and, perhaps, by a combined moderate enhancement of bile

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service