Skip to Content
Merck
All Photos(1)

Key Documents

BP100

Dacarbazine

British Pharmacopoeia (BP) Reference Standard

Synonym(s):

5-(3,3-Dimethyl-1-triazenyl)imidazole-4-carboxamide, DTIC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10N6O
CAS Number:
Molecular Weight:
182.18
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

dacarbazine

form

solid

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

BP

application(s)

pharmaceutical
pharmaceutical small molecule

format

neat

storage temp.

2-8°C

SMILES string

CN(C)\N=N\c1[nH]cnc1C(N)=O

InChI

1S/C6H10N6O/c1-12(2)11-10-6-4(5(7)13)8-3-9-6/h3H,1-2H3,(H2,7,13)(H,8,9)/b11-10+

InChI key

FDKXTQMXEQVLRF-ZHACJKMWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Dacarbazine BP Reference standard, intended for use in laboratory tests only as specifically prescribed in the British Pharmacopoeia.

Also used in monographs such as:
  • Dacarbazine Injection
  • Packaging

    Unit quantity: 25 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity please visit British Pharmacopoeia

    Other Notes

    Sales restrictions may apply.

    Pictograms

    Health hazardExclamation mark

    Signal Word

    Danger

    Hazard Classifications

    Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Eye Irrit. 2 - Muta. 1B - Skin Irrit. 2 - STOT SE 3

    Target Organs

    Respiratory system

    Storage Class Code

    6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

    WGK

    WGK 3

    Flash Point(F)

    Not applicable

    Flash Point(C)

    Not applicable


    Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

    If you need assistance, please contact Customer Support.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Ana Rita Ferreira et al.
    Membranes, 11(1) (2021-01-16)
    In the era of antibiotic resistance, there is an urgent need for efficient antibiotic therapies to fight bacterial infections. Cationic antimicrobial peptides (CAMP) are promising lead compounds given their membrane-targeted mechanism of action, and high affinity towards the anionic composition
    Junwei Hou et al.
    Nature cell biology, 22(10), 1264-1275 (2020-09-16)
    Although pyroptosis is critical for macrophages against pathogen infection, its role and mechanism in cancer cells remains unclear. PD-L1 has been detected in the nucleus, with unknown function. Here we show that PD-L1 switches TNFα-induced apoptosis to pyroptosis in cancer

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service