Skip to Content
Merck
All Photos(1)

Documents

BCR271

Benz[a]anthracene

BCR®, certified reference material

Synonym(s):

1,2-Benzanthracene, Tetraphene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H12
CAS Number:
Molecular Weight:
228.29
Beilstein:
1909298
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

437.6 °C (lit.)

mp

157-159 °C (lit.)

format

neat

storage temp.

2-8°C

SMILES string

c1ccc2cc3c(ccc4ccccc34)cc2c1

InChI

1S/C18H12/c1-2-7-15-12-18-16(11-14(15)6-1)10-9-13-5-3-4-8-17(13)18/h1-12H

InChI key

DXBHBZVCASKNBY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

For more information please see:
BCR271

Legal Information

BCR is a registered trademark of European Commission

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Atika Jaruchotikamol et al.
Toxicology and applied pharmacology, 224(2), 156-162 (2007-09-11)
The effects of andrographolide, the major diterpenoid constituent of Andrographis paniculata, on the expression of cytochrome P450 superfamily 1 members, including CYP1A1, CYP1A2, and CYP1B1, as well as on aryl hydrocarbon receptor (AhR) expression in primary cultures of mouse hepatocytes
T Kido et al.
Environmental toxicology, 28(1), 21-30 (2011-03-09)
Chlorinated benz[a]anthracenes (Cl-BaA) are halogenated aromatic compounds (typified by dioxins) found in the environment at relatively high concentrations. Fischer 344 rats were intragastrically administered 0, 1, or 10 mg of Cl-BaA or its parent compound benz[a]anthracene (BaA) per kg of
Maiysha D Jones et al.
Environmental microbiology, 13(10), 2623-2632 (2011-05-14)
The bacteria responsible for the degradation of naphthalene, phenanthrene, pyrene, fluoranthene or benz[a]anthracene in a polycyclic aromatic hydrocarbon (PAH)-contaminated soil were investigated by DNA-based stable-isotope probing (SIP). Clone libraries of 16S rRNA genes were generated from the (13) C-enriched ('heavy')
Sona Marvanová et al.
Chemical research in toxicology, 21(2), 503-512 (2008-01-22)
Monomethylated benz[ a]anthracenes (MeBaAs) are an important group of methylated derivatives of polycyclic aromatic hydrocarbons (PAHs). Although the methyl substitution reportedly affects their mutagenicity and tumor-initiating activity, little is known about the impact of methylation on the effects associated with
Yanzheng Gao et al.
Journal of hazardous materials, 172(2-3), 897-903 (2009-08-21)
Understanding the forms and availabilities of polycylic aromatic hydrocarbons (PAHs) would have considerable benefits for their risk assessment, and is of crucial importance for food security and remediation strategies in contaminated sites. In this work, the forms of six PAHs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service