Skip to Content
Merck
All Photos(1)

Key Documents

BCR138

Dibenz[a,h]anthracene

BCR®, certified reference material

Synonym(s):

1,2:5,6-Dibenzanthracene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H14
CAS Number:
Molecular Weight:
278.35
Beilstein:
1912416
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

524 °C (lit.)

mp

262-265 °C (lit.)

format

neat

storage temp.

2-8°C

SMILES string

c1ccc2c(c1)ccc3cc4c(ccc5ccccc45)cc23

InChI

1S/C22H14/c1-3-7-19-15(5-1)9-11-17-14-22-18(13-21(17)19)12-10-16-6-2-4-8-20(16)22/h1-14H

InChI key

LHRCREOYAASXPZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

For more information please see:
BCR138

Legal Information

BCR is a registered trademark of European Commission

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M F Kirby et al.
Aquatic toxicology (Amsterdam, Netherlands), 81(3), 233-244 (2007-01-24)
The extent to which biological systems interact in fish from multi-contaminant areas needs to be understood for full interpretation of monitoring data. This study investigates the interaction between two biomarkers, ethoxyresorufin-O-deethylase (EROD) activity and plasma vitellogenin (VTG) in the European
Santiago A Bortolato et al.
Analytical chemistry, 80(21), 8276-8286 (2008-10-02)
This work presents a novel approach for the simultaneous ultratrace determination of benzo[ a]pyrene and dibenzo[ a,h]anthracene, the two most carcinogenic polycyclic aromatic hydrocarbons (PAHs), in a very interfering environment, combining the recently discovered ability of the nylon membrane to
Katarzyna Skupinska et al.
Toxicology in vitro : an international journal published in association with BIBRA, 23(5), 763-771 (2009-04-14)
Polycyclic aromatic hydrocarbons (PAHs)--environmental carcinogens--are metabolized by CYP1A1 and CYP1A2 enzymes to oxy-derivatives, which are able to bind to DNA and initiate carcinogenesis. PAHs induce CYP1A1 and CYP1A2 activity, which increases the risk of development of carcinogenesis. Isothiocyanates (ITCs), naturally
Sander Delahaye et al.
Journal of chromatography. A, 1258, 152-160 (2012-09-04)
Supercritical fluid chromatography (SFC) is attributed many advantages over high performance liquid chromatography (HPLC). Next to the fact that SFC is greener than HPLC, which is especially important for preparative separations, SFC is claimed to be able to deliver faster
K B Okona-Mensah et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 43(7), 1103-1116 (2005-04-19)
Evidence suggests that people living in urban areas have an increased risk of lung cancer due to higher levels of air pollution in these areas. Benzo[a]pyrene (B[a]P) is currently used as the main indicator of carcinogenic polycyclic aromatic hydrocarbons (PAHs)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service