Skip to Content
Merck
All Photos(1)

Key Documents

B1160000

Bupivacaine hydrochloride

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H28N2O · HCl
CAS Number:
Molecular Weight:
324.89
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

bupivacaine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[Cl-].[N+H]1(C(CCCC1)C(=O)Nc2c(cccc2C)C)CCCC

InChI

1S/C18H28N2O.ClH/c1-4-5-12-20-13-7-6-11-16(20)18(21)19-17-14(2)9-8-10-15(17)3;/h8-10,16H,4-7,11-13H2,1-3H3,(H,19,21);1H

InChI key

SIEYLFHKZGLBNX-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Bupivacaine hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael R Fettiplace et al.
Anesthesiology, 120(4), 915-925 (2014-02-06)
Recent publications have questioned the validity of the "lipid sink" theory of lipid resuscitation while others have identified sink-independent effects and posed alternative mechanisms such as hemodilution. To address these issues, the authors tested the dose-dependent response to intravenous lipid
Qing-he Zhou et al.
Anesthesia and analgesia, 119(1), 203-206 (2014-05-09)
The spread of spinal anesthesia is highly unpredictable. In patients with increased abdominal girth and short stature, a greater cephalad spread after a fixed amount of subarachnoidally administered plain bupivacaine is often observed. We hypothesized that there is a strong
Mingquan Chen et al.
Anesthesia and analgesia, 118(4), 863-868 (2014-03-22)
In this study, we sought to determine the median effective dose (ED50) for motor block of intrathecally administered plain bupivacaine in adults (20-80 years) and to assess the effect of age on ED50 required for motor block. This study was
Ting Li et al.
International journal of molecular sciences, 20(17) (2019-09-06)
The G-protein-coupled receptor (GPCR) regulated intracellular signaling pathway is known to be involved in the development of insecticide resistance in the mosquito, Culex quinquefasciatus. To elucidate the specific role of each effector in the GPCR regulating pathway, we initially expressed
David Buck et al.
Anesthesia and analgesia, 119(1), 137-140 (2014-06-20)
An otherwise healthy 11-month-old, 8-kg infant presented for an elective circumcision. After a penile block with an excessive dose of 0.5% bupivacaine, the patient progressed to ventricular tachycardia. He was resuscitated with intralipid and had an uneventful recovery. The case

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service