Skip to Content
Merck
All Photos(1)

Key Documents

B1152000

Bromperidol

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

4-[4-(4-Bromophenyl)-4-hydroxy-1-piperidinyl]-1-(4-fluorophenyl)-1-butanone, R 11333

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H23BrFNO2
CAS Number:
Molecular Weight:
420.32
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

bromperidol

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

BrC(C=C1)=CC=C1C(CC2)(O)CCN2CCCC(C3=CC=C(F)C=C3)=O

InChI

1S/C21H23BrFNO2/c22-18-7-5-17(6-8-18)21(26)11-14-24(15-12-21)13-1-2-20(25)16-3-9-19(23)10-4-16/h3-10,26H,1-2,11-15H2

InChI key

RKLNONIVDFXQRX-UHFFFAOYSA-N

Gene Information

human ... DRD2(1813)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Bromperidol EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Butyrophenone antipsychotic; D2 dopamine and 5-HT2A serotonin antagonist.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Ohye et al.
Stereotactic and functional neurosurgery, 77(1-4), 33-39 (2002-10-16)
Recent advances on understanding the pallidothalamic relation lead us to perform Vim-Vo thalamotomy (combined thalamic lesion in ventralis intermedius nucleus and ventralis oralis nucleus) for cases with dyskinesia. In our recent series of thalamotomies, there are 12 cases of dyskinesia
Norio Yasui-Furukori et al.
Therapeutic drug monitoring, 26(3), 336-341 (2004-05-29)
The authors have developed and verified the precision and accuracy of new automated cloned enzyme donor immunoassays (CEDIA) for haloperidol and bromperidol, and cross-validations have been performed with conventional semiautomated EIA kits (MARKIT-M) and high-performance liquid chromatographic (HPLC) methods. The
Norio Yasui-Furukori et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 805(1), 175-180 (2004-04-29)
This study describes a new simultaneous determination of haloperidol and bromperidol and their reduced metabolites by modification of automated column-switching high-performance liquid chromatography. The test compounds were extracted from 1ml of plasma using chloroform-hexane (30:70 (v/v)), and the extract was
Lowijs N M Perquin
International clinical psychopharmacology, 20(6), 335-338 (2005-09-30)
Conventional antipsychotics are often associated with late-occurring undesirable movement effects. To gain more experience with newer antipsychotic agents, 151 patients in an acute admissions department were switched from treatment with conventional antipsychotics to sertindole between late 1996 and early 1998.
Norio Yasui-Furukori et al.
Therapeutic drug monitoring, 25(6), 709-714 (2003-11-26)
Therapeutic drug monitoring (TDM) services for plasma concentrations of haloperidol and bromperidol using enzyme immunoassay (EIA) methods are available in Japan, whereas high-performance liquid chromatographic (HPLC) methods are preferred in other countries. To compare these methods, we took 54 plasma

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service