Skip to Content
Merck
All Photos(2)

Documents

A8577

Sigma-Aldrich

Bovine Serum Albumin solution

30% in saline, protease free, aseptically filled

Synonym(s):

Albumin solution from bovine serum, BSA solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.27

biological source

bovine

Quality Level

sterility

aseptically filled

Assay

28-32% protein basis (biuret)

form

solution

purified by

heat shock fractionation

does not contain

stabilizer or preservative

origin

USA origin

concentration

28-32% protein (biuret)
30% in saline

technique(s)

ELISA: suitable
electrophoresis: suitable
western blot: suitable

UniProt accession no.

foreign activity

Alkaline phosphatase ≤0.001 units/mg protein
Peroxidase ≤0.001 units/mg protein
Protease ≤0.0001 units/mg protein

storage temp.

2-8°C

Gene Information

bovine ... ALB(280717)

Looking for similar products? Visit Product Comparison Guide

General description

Albumin is the most abundant plasma protein in humans. High levels of albumin are associated with dehydration. Albumin turnover is seen in infants with iron deficiency anemia. Serum albumin is a reliable prognostic indicator for liver disease. Oxidative damage of albumin is associated with advanced liver disease.

Application

Bovine Serum Albumin solution is suitable for use as a component in tris-buffered saline-triton-X-100 (TBS-T).

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michiko Harao et al.
Cancer immunology research, 5(6), 439-445 (2017-05-06)
Triple-negative breast cancer (TNBC) highly infiltrated with CD8
Oxidative damage of albumin in advanced liver disease
Oettl K, et al.
Biochimica et Biophysica Acta, 1782(7-8), 469-473 (2008)
Wei Xu et al.
Nature protocols, 16(8), 4084-4107 (2021-07-21)
Profiling chromatin accessibility at the single-cell level provides critical information about cell type composition and cell-to-cell variation within a complex tissue. Emerging techniques for the interrogation of chromatin accessibility in individual cells allow investigation of the fundamental mechanisms that lead
Chunxia Li et al.
Glycobiology, 31(8), 931-946 (2021-05-13)
Glycan microarrays have played important roles in detection and specificity assignment of glycan recognition by proteins. However, the size and diversity of glycan libraries in current microarray systems are small compared to estimated glycomes, and these may lead to missed
Expression of the GLP-1 receptor in mouse, rat, and human pancreas
Tornehave D, et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 56(9), 841-851 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service