Skip to Content
Merck
All Photos(1)

Key Documents

A0363000

Amantadine hydrochloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

1-Adamantanamine hydrochloride, 1-Adamantylamine hydrochloride, 1-Aminoadamantane hydrochloride, NSC 83653, Tricyclo[3.3.1.13,7]decan-1-amine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H17N · HCl
CAS Number:
Molecular Weight:
187.71
Beilstein:
4198854
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

amantadine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cl[H].[H][C@@]12C[C@@]3([H])C[C@@]([H])(C1)CC(N)(C2)C3

InChI

1S/C10H17N.ClH/c11-10-4-7-1-8(5-10)3-9(2-7)6-10;/h7-9H,1-6,11H2;1H/t7-,8+,9-,10-;

InChI key

WOLHOYHSEKDWQH-SOVZANNPSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Amantadine hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Dopamine releaser used to treat Parkinsonism and drug-induced extrapyramidal reactions.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amantadine hydrochloride: current and new uses.
S de Roin et al.
The Journal of neuroscience nursing : journal of the American Association of Neuroscience Nurses, 22(5), 322-325 (1990-10-01)
S Ito et al.
Nihon Kokyuki Gakkai zasshi = the journal of the Japanese Respiratory Society, 38(12), 897-902 (2001-03-14)
A virus infection was studied using half of the normal oral dose of amantadine hydrochloride-100 mg/d instead of 200 mg/d. The patients in this study, who visited the clinics during January and February 1999, were confirmed within 48 hours to
Bo OuYang et al.
Biochimica et biophysica acta, 1838(4), 1058-1067 (2013-09-24)
Many viral genomes encode small, integral membrane proteins that form homo-oligomeric channels in membrane, and they transport protons, cations, and other molecules across the membrane barrier to aid various steps of viral entry and maturation. These viral proteins, collectively named
R L Tominack et al.
Infectious disease clinics of North America, 1(2), 459-478 (1987-06-01)
Amantadine and rimantadine are oral antiviral drugs useful in the prophylaxis and treatment of influenza A virus infections. This article reviews the pharmacology, antiviral activity and mechanism of action, pharmacokinetics, toxicities, efficacy, and clinical applications of these agents. When administered
Matias Rey-Carrizo et al.
Journal of medicinal chemistry, 57(13), 5738-5747 (2014-06-19)
Amantadine inhibits the M2 proton channel of influenza A virus, yet most of the currently circulating strains of the virus carry mutations in the M2 protein that render the virus amantadine-resistant. While most of the research on novel amantadine analogues

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service