Skip to Content
Merck
All Photos(1)

Key Documents

A0300000

Adrenaline tartrate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(−)-Epinephrine (+)-bitartrate salt, L-3,4-Dihydroxy-α-(methylaminomethyl)benzyl alcohol D-hydrogen bitartrate salt, L-Adrenaline (+)-bitartrate salt, L-Adrenaline D-hydrogentartrate, L-Epinephrine D-hydrogentartrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H13NO3 · C4H6O6
CAS Number:
Molecular Weight:
333.29
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

adrenaline

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

O[C@H]([C@@H](O)C(O)=O)C(O)=O.CNC[C@H](O)c1ccc(O)c(O)c1

InChI

1S/C9H13NO3.C4H6O6/c1-10-5-9(13)6-2-3-7(11)8(12)4-6;5-1(3(7)8)2(6)4(9)10/h2-4,9-13H,5H2,1H3;1-2,5-6H,(H,7,8)(H,9,10)/t9-;1-,2-/m01/s1

InChI key

YLXIPWWIOISBDD-NDAAPVSOSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Adrenaline tartrate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Adrenergic receptor agonist.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 1 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nandini Pathak et al.
3 Biotech, 10(10), 438-438 (2020-10-01)
Considering the cognitive impairment induced by temozolomide (TMZ) in glioblastoma survivors, the present study was aimed to evaluate the protective effect of dehydrozingerone (DHZ) against TMZ-induced cognitive impairment (chemobrain) and C6 cell line-induced glioma in male Wistar rats. In both
Routine determination of plasma catecholamines using reversed-phase, ion-pair high-performance liquid chromatography with electrochemical detection.
DaviesL C and Molyneux G S.
Journal of Chromatography. B, Biomedical Sciences and Applications, 231(1), 41-51 (1982)
Jun Pu et al.
Biochemical and biophysical research communications, 493(3), 1273-1279 (2017-10-03)
Psychological stress has recently been described as a risk factor in the development of pancreatic cancer. Here, we reported that increased neurotransmitter adrenaline was associated with the poor survival in pancreatic cancer patients. Moreover, in the cell model study, we
Filipa Amaro et al.
International journal of molecular sciences, 21(21) (2020-10-31)
Adrenaline, which participates in the neuroendocrine response that occurs during stress and perimenopause, may be tumorigenic. This exploratory study aimed at investigating whether non-tumorigenic and tumorigenic human breast epithelial cell lines are able to synthesize adrenaline. The study was carried
Sanatan Mishra et al.
Life sciences, 218, 96-111 (2018-12-24)
Our earlier studies revealed the cardio-protective effects of oleic acid, a monounsaturated fatty acid, against adrenaline induced myocardial injury. Moreover, it has been found to possess antioxidant properties. Thus, in the present study we have investigated the protective role of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service