Skip to Content
Merck
All Photos(1)

Key Documents

97326

Supelco

Isosilybin A

analytical standard

Synonym(s):

(2R,3R)-3,5,7-Trihydroxy-2-[(2R,3R)-2-(4-hydroxy-3-methoxyphenyl)-3-(hydroxymethyl)-2,3-dihydrobenzo[1,4]dioxin-6-yl]-4-chromanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H22O10
CAS Number:
Molecular Weight:
482.44
UNSPSC Code:
85151701
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤10.0% water

application(s)

food and beverages

format

neat

SMILES string

COc1cc(ccc1O)[C@H]2Oc3ccc(cc3O[C@@H]2CO)[C@H]4Oc5cc(O)cc(O)c5C(=O)[C@@H]4O

InChI

1S/C25H22O10/c1-32-17-6-11(2-4-14(17)28)24-20(10-26)33-18-7-12(3-5-16(18)34-24)25-23(31)22(30)21-15(29)8-13(27)9-19(21)35-25/h2-9,20,23-29,31H,10H2,1H3/t20-,23+,24-,25-/m1/s1

InChI key

FDQAOULAVFHKBX-HKTJVKLFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Isosilybin A is a flavonolignan and one of the bioactive component available in silymarin, which is isolated from Silybum marianum. It is commonly used in the treatment of cirrhosis, hepatitis and prevents the liver from being exposed to toxic substances.

Application

Isosilybin B may be used as an analytical reference standard for the determination of the analyte in compound preparations of Silybum marianum and Silybum miltiorrhiza using high-performance liquid chromatography with photo diode array detection and electrospray ionization-mass spectrometry.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Simultaneous determination of abietane-type diterpenes, flavonolignans and phenolic compounds in compound preparations of Silybum marianum and Salvia miltiorrhiza by HPLC-DAD-ESI MS
Zhao Y, et al.
Journal of Pharmaceutical and Biomedical Analysis, 38(3), 564-570 (2005)
Separation and characterization of silybin, isosilybin, silydianin and silychristin in milk thistle extract by liquid chromatography?electrospray tandem mass spectrometry
Lee.IJ, et al.
Journal of Chromatography A, 1116(1-2), 147-152 (2006)
Konstantina Filippopoulou et al.
Free radical biology & medicine, 103, 256-267 (2017-01-01)
Aging is an unavoidable process characterized by gradual failure of homeostasis that constitutes a critical risk factor for several age-related disorders. It has been unveiled that manipulation of various key pathways may decelerate the aging progression and the triggering of
Jiazeng Yang et al.
Metabolic engineering, 59, 44-52 (2020-02-01)
Silymarin extracted from milk thistle seeds, is used for treating hepatic diseases. Silybin and isosilybin are its main components, and synthesized from coupling of taxifolin and coniferyl alcohol. Here, the biosynthetic pathways of taxifolin and coniferyl alcohol were reconstructed in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service