Skip to Content
Merck
All Photos(2)

Key Documents

93632

Supelco

Zinc oxide

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZnO
CAS Number:
Molecular Weight:
81.39
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Assay

≥99.0% (KT)

shelf life

limited shelf life, expiry date on the label

technique(s)

AAS: suitable
HPLC: suitable
ICP: suitable
gas chromatography (GC): suitable

cation traces

As: ≤2 mg/kg
Cd: ≤10 mg/kg
Fe: ≤10 mg/kg
Pb: ≤10 mg/kg

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

O=[Zn]

InChI

1S/O.Zn

InChI key

XLOMVQKBTHCTTD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food.

Find all available reference materials for compounds listed in 10/2011 here

Application

Employed in the preparation of NaZnSiO3OH, a novel chiral framework material which has potential application in ion exchange, adsorption or catalysis.
Zinc oxide has been used as a raw material in the synthesis of environmental friendly silver front contact paste comprising of silver particles, glass frit powders, and resin binder, which can be used in crystalline silicon solar cells.[1]

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A new environmental friendly silver front contact paste for crystalline silicon solar cells
Che Q, et al.
J. Alloy Compounds, 549, 221-225 (2013)
Cui Li et al.
Annals of botany, 123(1), 57-68 (2018-07-19)
The pathways whereby foliar-applied nutrients move across the leaf surface remain unclear. The aim of the present study was to examine the pathways by which foliar-applied Zn moves across the sunflower (Helianthus annuus) leaf surface, considering the potential importance of
Chia-Chi Wang et al.
Journal of nanoscience and nanotechnology, 13(6), 3880-3888 (2013-07-19)
Zinc oxide nanoparticles (nano-ZnO) are one of the most commonly used nanomaterials in industrial products including paints, cosmetics, and medical materials. Since ZnO is a well-known photocatalyst, it is important to further study if nano-ZnO cause phototoxic effect on skin
Jeongmin Kang et al.
Journal of nanoscience and nanotechnology, 13(5), 3526-3528 (2013-07-19)
ZnO-nanowire-based logic circuits were constructed by the vertical integration of multilayered field-effect transistors (FETs) on plastic substrates. ZnO nanowires with an average diameter of -100 nm were synthesized by thermal chemical vapor deposition for use as the channel material in
Dadong Guo et al.
Journal of nanoscience and nanotechnology, 13(6), 3769-3777 (2013-07-19)
Nanomaterials, including zinc oxide (ZnO) nanoparticles, are being developed for a variety of commercial products. Recent reports showed that cells exposed to ZnO nanoparticles produced severe cytotoxicity accompanied by oxidative stress and genotoxicity. To understand the possible mechanism underlying oxidative

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service