Skip to Content
Merck
All Photos(2)

Key Documents

91466

Supelco

Betulinic acid

analytical standard

Synonym(s):

3β-Hydroxy-20(29)-lupaene-28-oic acid, Lupatic acid, Mairin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H48O3
CAS Number:
Molecular Weight:
456.70
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

295-298 °C (dec.) (lit.)

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

CC(=C)[C@@H]1CC[C@@]2(CC[C@]3(C)[C@H](CC[C@@H]4[C@@]5(C)CC[C@H](O)C(C)(C)[C@@H]5CC[C@@]34C)[C@@H]12)C(O)=O

InChI

1S/C30H48O3/c1-18(2)19-10-15-30(25(32)33)17-16-28(6)20(24(19)30)8-9-22-27(5)13-12-23(31)26(3,4)21(27)11-14-29(22,28)7/h19-24,31H,1,8-17H2,2-7H3,(H,32,33)/t19-,20+,21-,22+,23-,24+,27-,28+,29+,30-/m0/s1

InChI key

QGJZLNKBHJESQX-FZFNOLFKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Betulinic acid is a pentacyclic triterpene, isolated from the leaves of Syzigium claviflorum. It is found to be an inhibitor of HIV replication in H9 lymphocyte cells.

Application

Betulinic acid may be used as a reference standard in the determination of betulinic acid in white birch bark using reversed-phase high-performance liquid chromatography (RP-HPLC).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Betulinic acid, a pentacyclic triterpene, selectively induces apoptosis in tumor cells by directly activating the mitochondrial pathway of apoptosis through a p53- and CD95-independent mechanism.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

This compound is commonly found in plants of the genus: glycyrrhiza melissa

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Simultaneous determination of betulin and betulinic acid in white birch bark using RP-HPLC.
Zhao G, et al.
Journal of Pharmaceutical and Biomedical Analysis, 43(3), 959-962 (2007)
Anti-AIDS agents, 11. Betulinic acid and platanic acid as anti-HIV principles from Syzigium claviflorum, and the anti-HIV activity of structurally related triterpenoids.
Fujioka T, et al.
Journal of Natural Products, 57(2), 243-247 (1994)
Discovery of betulinic acid as a selective inhibitor of human melanoma that functions by induction of apoptosis.
Pisha E, et al.
Nature Medicine, 1(10), 1046-1046 (1995)
Christopher Aiken et al.
Trends in molecular medicine, 11(1), 31-36 (2005-01-15)
Betulinic acid (BA) derivatives are low molecular weight organic compounds synthesized from a plant-derived natural product. Several BA derivatives are potent and highly selective inhibitors of HIV-1. Depending on the specific side-chain modification, these compounds function by inhibiting HIV fusion
Yan-Fang Sun et al.
Food chemistry, 138(2-3), 1998-2007 (2013-02-16)
Betulinic acid (BA), a natural pentacyclic triterpenoid, was isolated from sour jujube fruits for the first time. An inclusion complex comprising BA and β-cyclodextrin (β-CD) was formed to improve the dissolution of BA, but little is known about its anticancer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service