Skip to Content
Merck
All Photos(1)

Key Documents

91446

Supelco

Pentadecanoic acid

analytical standard

Synonym(s):

C15:0, NSC 28486, Pentadecylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)13COOH
CAS Number:
Molecular Weight:
242.40
Beilstein:
1773831
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

257 °C/100 mmHg (lit.)

mp

51-53 °C (lit.)
51-55 °C

format

neat

functional group

carboxylic acid

shipped in

ambient

storage temp.

room temp

SMILES string

CCCCCCCCCCCCCCC(O)=O

InChI

1S/C15H30O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15(16)17/h2-14H2,1H3,(H,16,17)

InChI key

WQEPLUUGTLDZJY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pentadecanoic acid is a saturated fatty acid, commonly present in ox bile. It is present as a phytochemical component of Indigofera suffruticosa leaves and is known to exhibit antimicrobial and antioxidant properties.

Application

Pentadecanoic acid has been used as an internal standard in the separation of chloroplast lipids like digalactosyldiacylglycerol, sulfoquinovosyldiacylglycerol and monogalactosyldiacylglycerol extracted from cyanobacterial cells and spotted on membranes using thin layer chromatography technique (TLC).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation of Chloroplast Lipid Membrane and Lipid-Protein Interaction Assay
Okazaki K, et al.
Bio-protocol, 6 (2016)
Identification by GC-FID and GC-MS of amino acids, fatty and bile acids in binding media used in works of art
Mateo-Castro.R, et al.
Fresenius Journal of Analytical Chemistry, 369(7-8), 642-646 (2001)
GC-MS analysis of bioactive constituents of Indigofera suffruticosa leaves
Vijisaral Elezabeth, D and S. Arumugam
Journal of Chemical and Pharmaceutical Research, 6(8), 294-300 (2014)
Shinro Matsuo et al.
Internal medicine (Tokyo, Japan), 43(6), 490-492 (2004-07-31)
A 72-year-old woman had insidious onset of heart failure, and was diagnosed as multicentric Castleman's disease. She underwent myocardial imaging with technetium-99m tetrofosmin, 1-123 beta-methyl-iodophenyl pentadecanoic acid (BMIPP). Technetium-99m tetrofosmin studies showed almost normal uptake of the left ventricular myocardium
Hidenori Seki et al.
Circulation journal : official journal of the Japanese Circulation Society, 69(3), 311-319 (2005-02-26)
(18)F-fluorodeoxyglucose (FDG) positron-emission tomography (PET) is assumed to be the most useful method of evaluating the viability of the myocardium, but its use is limited by the need for a cyclotron. In the present study, the ability of a combination

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service