Skip to Content
Merck
All Photos(1)

Key Documents

91301

Supelco

Dichloromethane

Selectophore, ≥99.5% (GC), inhibitor-free

Synonym(s):

Methylene chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH2Cl2
CAS Number:
Molecular Weight:
84.93
Beilstein:
1730800
EC Number:
MDL number:
UNSPSC Code:
12191501
PubChem Substance ID:
NACRES:
NB.61
grade:
for ion-selective electrodes
technique(s):
GC/GC: suitable
bp:
39.8-40 °C (lit.)
vapor pressure:
24.45 psi ( 55 °C)
6.83 psi ( 20 °C)

grade

for ion-selective electrodes

Quality Level

vapor density

2.9 (vs air)

vapor pressure

24.45 psi ( 55 °C)
6.83 psi ( 20 °C)

product line

Selectophore

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

1223 °F

expl. lim.

22 %

technique(s)

GC/GC: suitable

impurities

≤0.02% water

evapn. residue

≤0.002%

refractive index

n20/D 1.423-1.425
n20/D 1.424 (lit.)

bp

39.8-40 °C (lit.)

mp

−95 °C (lit.)

density

1.325 g/mL at 25 °C (lit.)

SMILES string

ClCCl

InChI

1S/CH2Cl2/c2-1-3/h1H2

InChI key

YMWUJEATGCHHMB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dichloromethane is a volatile halogenated organic solvent used as a degreaser, extraction medium, lacquer remover, or paint stripper.

Application

Dichloromethane may be used as a reagent for the extraction of lipophilic anions across a carrier-mediated polymer inclusion membrane, driven by an electric field.

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Accidental intoxication by dichloromethane at work place: Clinical case and literature review
Pacheco C, et al.
Journal of Acute Medicine, 6(2), 43-45 (2016)
Dichloromethane-degrading bacteria in the genomic age.
Muller E, et al.
Research in Microbiology, 162(9), 869-876 (2011)
Hong Heng See et al.
Analytical chemistry, 83(19), 7507-7513 (2011-08-20)
The use of a cationic carrier-mediated polymer inclusion membrane (PIM) for extraction and preconcentration of anionic model analytes driven by an electric field directly into an aqueous acceptor solution is demonstrated. The optimized membrane was 20 μm thick and consisted

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service