Skip to Content
Merck
All Photos(4)

Key Documents

902179

Sigma-Aldrich

Calcium chloride

anhydrous, granular, ≤7.0 mm, free-flowing, Redi-Dri, ReagentPlus®, ≥93%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CaCl2
CAS Number:
Molecular Weight:
110.98
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.55
grade:
anhydrous
form:
granular

grade

anhydrous

vapor pressure

0.01 mmHg ( 20 °C)

product line

ReagentPlus®
Redi-Dri

Assay

≥93%

form

granular

quality

free-flowing

particle size

≤7.0 mm

mp

772 °C (lit.)

SMILES string

[Cl-].[Cl-].[Ca++]

InChI

1S/Ca.2ClH/h;2*1H/q+2;;/p-2

InChI key

UXVMQQNJUSDDNG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Calcium chloride was used during the isolation of total β-glucan from hydrolyzed barley powders. It was also used as a cross-linker during the preparation of Aloe vera pectin gels.
It may be used in the following processes:
  • Modification of natural zeolite to increase its ability of fluoride removal.
  • Synthesis of calcium sulfate whiskers utilized in making polymer reinforcing composite materials.
  • Synthesis of calcium chloride-anodized aluminum composites, a solid sorbent.
  • As an extractant in the soil analysis process.
  • As a catalyst in the synthesis of aldols by reacting dimethylsilyl (DMS) enolates with aldehydes.

Caution

may contain powder and some dark grey granules

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany
Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sang Hoon Lee et al.
Preventive nutrition and food science, 20(2), 110-118 (2015-07-16)
This study was performed to investigate changes in the content and purity, as well as physical characteristics of β-glucan extracted from acid hydrolyzed whole grain barleys. Waxy and non-waxy barleys (Hordeum vulgare) were hydrolyzed with different concentrations of HCl (0.1~0.5
Pectins from Aloe Vera: Extraction and production of gels for regenerative medicine.
Gentilini R, et al.
Journal of Applied Polymer Science, 131(2) (2014)
Ángel Serrano-Aroca et al.
PloS one, 12(9), e0185235-e0185235 (2017-09-22)
Micrometer length tubes of graphene oxide (GO) with irregular form were synthesised following facile and green metal complexation reactions. These materials were obtained by crosslinking of GO with calcium, zinc or strontium chlorides at three different temperatures (24, 34 and
Skin and soft tissue necrosis from calcium chloride in a deicer.
Kim MP, et al.
The Journal of Emergency Medicine, 32(1), 41-44 (2007)
A review on the use of calcium chloride in applied thermal engineering.
N'Tsoukpoe KE, et al.
Applied Thermal Engineering, 75, 513-531 (2015)

Articles

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service