Skip to Content
Merck
All Photos(1)

Documents

88413

Supelco

1-(2-Thiazolylazo)-2-naphthol

for spectrophotometric det. of Cd, Co, Cu, Mn, Ni, Tl, ≥99.0%

Synonym(s):

2-(2-Hydroxy-1-naphthylazo)thiazole, TAN

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H9N3OS
CAS Number:
Molecular Weight:
255.30
Beilstein:
21172
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.21

Quality Level

Assay

≥99.0% (CHN)
≥99.0%

form

powder

quality

for spectrophotometric det. of Cd, Co, Cu, Mn, Ni, Tl

technique(s)

UV/Vis spectroscopy: suitable

mp

138-139 °C

solubility

methanol: 10 mg/mL, clear

SMILES string

Oc1ccc2ccccc2c1\N=N\c3nccs3

InChI

1S/C13H9N3OS/c17-11-6-5-9-3-1-2-4-10(9)12(11)15-16-13-14-7-8-18-13/h1-8,17H/b16-15+

InChI key

IOMXCGDXEUDZAK-FOCLMDBBSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Reagent for the extraction and spectrophotometric determination of metals, metallochromic indicator

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Serife Tokalioğlu et al.
Environmental monitoring and assessment, 152(1-4), 369-377 (2008-05-20)
A new method for separation and preconcentration of trace amounts of Cu(II), Ni(II), Pb(II), Cd(II) and Mn(II) ions in various matrices was proposed. The method is based on the adsorption and chelation of the metal ions on a column containing
Y Sumi et al.
Histochemistry, 87(4), 327-329 (1987-01-01)
We describe a new autoradiographic technique for differentially demonstrating the presence of trace amounts of cadmium that remains unbound to metallothionein in tissue sections. This procedure involves [14C-1,4,5,8-naphthol]-1-(2-thiazolylazo)-2-naphthol (TAN) instead of a metal radioisotope. Tissue sections were stained using labeled
K C Teo et al.
The Analyst, 126(4), 534-537 (2001-05-09)
Cloud point extraction has been used for the preconcentration of manganese, after the formation of a complex with 1-(2-thiazolylazo)-2-naphthol (TAN), and later analysis by flame atomic absorption spectrometry using octylphenoxypolyethoxyethanol (Triton X-114) as surfactant. The chemical variables affecting the separation
Z. Holzbecher et al.
Handbook of Organic Reagents in Inorganic Analysis (1976)
M M Omar et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 61(5), 929-936 (2005-02-03)
The synthesis and characterization of Mn(II), Fe(III), Co(II), Ni(II), Cu(II), Zn(II), Cd(II), Pd(II) and UO2(II) chelates of 1-(2-thiazolylazo)-2-naphthalenol (TAN) were reported. The dissociation constants of the ligand and the stability constants of the metal complexes were calculated pH-metrically at 25

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service