Skip to Content
Merck
All Photos(1)

Key Documents

86959

Sigma-Aldrich

Clara-Diastase

mixture of enzymes, powder, ≥35 U/mg

Synonym(s):

Clarase 300

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.21

Quality Level

form

powder

specific activity

≥35 U/mg

storage condition

under inert gas

color

beige

solubility

soluble

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Clara-Diastase is a mixture of α-amylase, cellulase, invertase, peptidase, phosphatase, and sulfatase. It is useful in efficient enzymatic hydrolysis.

Unit Definition

1 U corresponds to the amount of enzyme which liberates 1 μmol maltose per minute at pH 6.0 and 25 °C (starch according to Zulkowsky, Cat. No. 85642, as substrate)

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Valls et al.
Journal of agricultural and food chemistry, 47(1), 170-173 (1999-11-24)
A reliable and rapid high-performance liquid chromatography (HPLC) method has been set up for the determination of total thiamin in difficult sample matrices such as cooked sausages. Different hydrolysis conditions and enzymes were tested to release the vitamin from its
Vaida Damulienė et al.
Foods (Basel, Switzerland), 12(19) (2023-10-14)
Bee pollen is one of the most valuable apitherapeutic products with high nutritional value. To obtain a higher diversity of compounds, higher bioactivity, and improve the release of nutrients from bee pollen, additional processing of the raw material may be

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service