Skip to Content
Merck
All Photos(1)

Key Documents

86440

Sigma-Aldrich

Dimethyl terephthalate

purum, ≥99.0% (GC)

Synonym(s):

DMT, Dimethyl 1,4-benzenedicarboxylate, Dimethyl p-benzenedicarboxylate, Dimethyl p-phthalate, Methyl 4-(carbomethoxy)benzoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,4-(CO2CH3)2
CAS Number:
Molecular Weight:
194.18
Beilstein:
1107185
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

1.04 (vs air)

Quality Level

vapor pressure

1.15 mmHg ( 93 °C)

grade

purum

Assay

≥99.0% (GC)

form

powder, crystals or chunks

autoignition temp.

1058 °F

mp

139-141 °C (lit.)
140-142 °C

SMILES string

COC(=O)c1ccc(cc1)C(=O)OC

InChI

1S/C10H10O4/c1-13-9(11)7-3-5-8(6-4-7)10(12)14-2/h3-6H,1-2H3

InChI key

WOZVHXUHUFLZGK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

309.2 °F - (External MSDS)

Flash Point(C)

154 °C - (External MSDS)

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Z Chen et al.
Guang pu xue yu guang pu fen xi = Guang pu, 21(3), 362-365 (2003-09-02)
The fluorescence quenching of perylene and pyrene by dimethyl terephthalate (DMTP) and N,N-dimethylaniline (DMA) have been investigated. The results show that pyrene can form exciplex with DMTP or DMA at room temperature and perylene can only form exciplex with DMA.
L Tserovska et al.
Acta microbiologica Bulgarica, 30, 67-71 (1993-01-01)
The mixed microbial culture 189 possesses high biodegradation capacity against dimethylterepthalate (DMT)--a basic substance in polyester fibres production and an environmental pollutant. The process of its degradation was followed and the effect of some external factors (pH, concentration, accompanying substrates)
R I Goncharova et al.
Mutation research, 204(4), 703-709 (1988-04-01)
The mutagenic activity of dimethyl terephthalate (DMtP) was evaluated in the micronucleus test in mice. A clear clastogenic effect was obtained at all concentrations studied (0.2-1.0 mmole/kg body weight). The maximum number of micronuclei occurred 24 h after a single
[The basophil degranulation reaction as a method for detecting allergy and autoallergy to common chemical compounds].
G I Vinogradov et al.
Laboratornoe delo, (6)(6), 339-341 (1984-01-01)
Yu Ping Wang et al.
Ecotoxicology (London, England), 15(6), 549-557 (2006-09-07)
Two strains of bacteria were isolated from deep-ocean sediments of the South China Sea using enrichment culturing technique and they were identified as Sphingomonas yanoikuyae DOS01 (AY878409) and Variovorax paradoxus T4 (AY878410) based on 16S rRNA gene sequences. S. yanoikuyae

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service