Skip to Content
Merck
All Photos(1)

Documents

84332

Sigma-Aldrich

Methyl salicylate

tested according to Ph. Eur.

Synonym(s):

Methylis salicylas, 2-Hydroxybenzoic acid methyl ester, Methyl 2-hydroxybenzoate, Oil of wintergreen, Wintergreen oil

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-(HO)C6H4CO2CH3
CAS Number:
Molecular Weight:
152.15
Beilstein:
971516
EC Number:
MDL number:
UNSPSC Code:
41116100
PubChem Substance ID:
NACRES:
NA.21

Agency

USP/NF
tested according to Ph. Eur.

Quality Level

vapor density

5.26 (vs air)

vapor pressure

1 mmHg ( 54 °C)

form

liquid

autoignition temp.

847 °F

refractive index

n20/D 1.536 (lit.)

bp

222 °C (lit.)

mp

−8-−7 °C (lit.)

density

1.174 g/mL at 25 °C (lit.)

SMILES string

COC(=O)c1ccccc1O

InChI

1S/C8H8O3/c1-11-8(10)6-4-2-3-5-7(6)9/h2-5,9H,1H3

InChI key

OSWPMRLSEDHDFF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Repr. 2 - Skin Sens. 1B

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

204.8 °F - closed cup

Flash Point(C)

96 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sven Hildebrand et al.
Scientific reports, 9(1), 10880-10880 (2019-07-28)
Optical clearing techniques and light sheet microscopy have transformed fluorescent imaging of rodent brains, and have provided a crucial alternative to traditional confocal or bright field techniques for thin sections. However, clearing and labeling human brain tissue through all cortical
Michiel van Wijk et al.
PloS one, 6(7), e21742-e21742 (2011-07-19)
Insect herbivory induces plant odors that attract herbivores' natural enemies. Assuming this attraction emerges from individual compounds, genetic control over odor emission of crops may provide a rationale for manipulating the distribution of predators used for pest control. However, studies
Nadir Erbilgin et al.
Tree physiology, 32(8), 946-957 (2012-06-05)
Coniferous trees have both constitutive and inducible defences that deter or kill herbivores and pathogens. We investigated constitutive and induced monoterpene responses of jack pine (Pinus banksiana Lamb.) to a number of damage types: a fungal associate of the mountain
Hsu Shao Feng et al.
Journal of clinical periodontology, 38(7), 637-643 (2011-04-01)
To evaluate the clinical efficacy of subgingival ultrasonic instrumentation irrigated with essential oils (EOs) of residual periodontal pockets. Sixty-four individuals with chronic periodontitis were invited to participate in this randomized, double-blind, parallel, and placebo-controlled clinical trial. All subjects received non-surgical
Prisca S Pierre et al.
Journal of chemical ecology, 37(4), 368-377 (2011-03-31)
Plants attacked by herbivorous insects emit volatile organic compounds that are used by natural enemies to locate their host or prey. The composition of the blend is often complex and specific. It may vary qualitatively and quantitatively according to plant

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service