Skip to Content
Merck
All Photos(1)

Key Documents

82270

Sigma-Aldrich

1,2-Dichloropropane

99%

Synonym(s):

Propylene dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CHClCH2Cl
CAS Number:
Molecular Weight:
112.99
Beilstein:
1718880
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

3.89 (vs air)

vapor pressure

40 mmHg ( 19.4 °C)

Assay

99%

form

liquid

autoignition temp.

1035 °F

expl. lim.

14.5 %

refractive index

n20/D 1.439 (lit.)
n20/D 1.439

bp

95-96 °C (lit.)

mp

−100 °C (lit.)

density

1.156 g/mL at 25 °C (lit.)

functional group

chloro

SMILES string

CC(Cl)CCl

InChI

1S/C3H6Cl2/c1-3(5)2-4/h3H,2H2,1H3

InChI key

KNKRKFALVUDBJE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,2-Dichloropropane, also known as Propylene dichloride, is a chlorinated solvent, that is commonly utilized as an intermediate in the synthesis of basic organic chemicals.

Application

1,2-Dichloropropane can be used as a building block to synthesize:
  •  2-Ylidene-1,3-dithiolane derivatives via one-pot reaction with carbon disulfide and active methylene compounds in the presence of sodium ethylate.      
  • β-Chloropropylsulfurs by controllable dechloro-coupling reaction with thiols.      
  • Thiazolidine and 1,3-thiazinane derivatives via [3+2] and [3+3] annulation reaction with β-ketothioamides in the presence of phosphonium ylide as a catalyst.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

59.0 °F - closed cup

Flash Point(C)

15.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of some 2-ylidene-1, 3-dithiolanes
Lipin KV, et al.
Russ. J. Org. Chem., 53(1), 147-149 (2017)
Monish Arbaz Ansari et al.
Organic & biomolecular chemistry, 17(41), 9151-9162 (2019-10-09)
Phosphonium ylides are being reported here as a catalyst for the formation of thiazolidines and 1,3-thiazinanes from β-ketothioamides (which act as a three atom N, C, and S synthon) with dihaloalkanes via [3 + 2] and [3 + 3] annulations
Divergent Electrolysis for the Controllable Coupling of Thiols with 1, 2-Dichloroethane: a Mild Approach to Sulfide and Sulfoxide
Ling F, et al.
Green Chemistry (2022)
J C Hage et al.
Applied microbiology and biotechnology, 57(4), 548-554 (2002-01-05)
Pseudomonas sp. strain DCA1, which is capable of utilizing 1,2-dichloroethane (DCA) as sole carbon and energy source, was used to oxidize chlorinated methanes, ethanes, propanes, and ethenes. Chloroacetic acid, an intermediate in the DCA degradation pathway of strain DCA1, was
A Trevisan et al.
Archives of toxicology, 66(9), 641-645 (1992-01-01)
Sex- and age-related nephrotoxicity due to 1,2-dichloropropane was studied in vitro by means of renal cortical slices obtained from Wistar rats. Reduced glutathione content, organic anion accumulation (p-aminohippurate), and release of malondialdehyde (to measure the extent of lipid peroxidation), aspartate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service