Skip to Content
Merck
All Photos(1)

Key Documents

79311

Supelco

Malvidin-3-galactoside chloride

analytical standard

Synonym(s):

Arthanitin chloride, Primulin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H25ClO12
CAS Number:
Molecular Weight:
528.89
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.25

grade

analytical standard

Quality Level

Assay

≥95% (HPLC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

[Cl-].COc1cc(cc(OC)c1O)-c2[o+]c3cc(O)cc(O)c3cc2O[C@@H]4O[C@H](CO)[C@H](O)[C@H](O)[C@H]4O

InChI

1S/C23H24O12.ClH/c1-31-14-3-9(4-15(32-2)18(14)27)22-16(7-11-12(26)5-10(25)6-13(11)33-22)34-23-21(30)20(29)19(28)17(8-24)35-23;/h3-7,17,19-21,23-24,28-30H,8H2,1-2H3,(H2-,25,26,27);1H/t17-,19+,20+,21-,23-;/m1./s1

InChI key

YDIKCZBMBPOGFT-PWUSVEHZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Malvidin-3-galactoside chloride is an anthocyanin pigment found in magenta and dark red flowers of Primula polyanthus.

Application

Malvidin-3-galactoside chloride is a type of fluorescent dye commonly used to identify glycerides and free fatty acids.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

S Trumbeckaitė et al.
Phytotherapy research : PTR, 27(4), 499-506 (2012-05-26)
In this study, the direct influence of bilberry (Vaccinium myrtillus) fruit extracts (aqueous and ethanolic) rich in anthocyanins on the oxidative phosphorylation of isolated rat heart mitochondria was investigated in vitro. Higher concentrations of bilberry extracts concentration-dependently inhibited mitochondrial state
V A Otellin et al.
Arkhiv anatomii, gistologii i embriologii, 80(1), 26-29 (1981-01-01)
A possibility to study associative and projective connections of the optic cortical fields by the method of luminescent revealing of primuline retrograde transport has been demonstrated in the experiments with cats. Initial neurons of the connective systems have been revealed
Rose Scott-Moncrieff and the dawn of (bio)chemical genetics.
Martin C.
The Biochemical Journal, 38(2), 48-53 (2016)
[Role of membrane damage in the inactivation of yeast cells by visible light].
M E Pospelov et al.
Mikrobiologiia, 56(5), 882-885 (1987-09-01)
P V Sarthy et al.
Neuroscience letters, 25(3), 205-208 (1981-09-25)
Mouse retinal ganglion cells were labeled by retrograde axonal transport of the fluorescent dyes, DAPI and Primulin. The labeled retinae were dispersed by papain treatment and mechanical dissociation. Among the cells in the suspension, the ganglion cells could be unequivocally

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service