Skip to Content
Merck
All Photos(1)

Key Documents

77619

Supelco

Phenol – chloroform – isoamyl alcohol mixture

Phenol – chloroform – isoamyl alcohol mixture

BioUltra, for molecular biology, 125:24:1

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
ND.02
grade:
for molecular biology
Pricing and availability is not currently available.

grade

for molecular biology

Quality Level

product line

BioUltra

form

liquid

impurities

DNases, none detected
RNases, none detected
phosphatases, none detected
proteases, none detected

pH

4-5(H2O-phase, after extraction with H2O, 1:1)

density

1.12 g/mL at 20 °C

λ

neat

UV absorption

λ: 330 nm Amax: 0.2
λ: 405 nm Amax: 0.1
λ: 510 nm Amax: 0.1

storage temp.

2-8°C

SMILES string

ClC(Cl)Cl.CC(C)CCO.Oc1ccccc1

InChI

1S/C6H6O.C5H12O.CHCl3/c7-6-4-2-1-3-5-6;1-5(2)3-4-6;2-1(3)4/h1-5,7H;5-6H,3-4H2,1-2H3;1H

InChI key

ZYWFEOZQIUMEGL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Simultaneous quantification of multiple RNA cargos co-loaded into nanoparticle-based delivery systems: This research explores the application of the Phenol – chloroform – isoamyl alcohol mixture in extracting high-quality RNA for advanced therapeutic formulations, emphasizing its critical role in nanotechnology and pharmaceutical sciences (Lokras et al., 2022).

  • Analysis of Circulating microRNAs and Their Post-Transcriptional Modifications in Cancer Serum by On-Line Solid-Phase Extraction-Capillary Electrophoresis-Mass Spectrometry: Demonstrates the use of the Phenol – chloroform – isoamyl alcohol mixture for precise extraction of microRNAs from biological fluids, contributing to cancer diagnostics and molecular pathology (Pero-Gascon et al., 2018).

  • The application of alkaline lysis and pressure cycling technology in the differential extraction of DNA from sperm and epithelial cells recovered from cotton swabs: Highlights the integration of the Phenol – chloroform – isoamyl alcohol mixture with novel technologies for forensic applications, enhancing DNA extraction processes critical in legal and criminal investigations (Nori and McCord, 2015).

  • Frequency of Epstein-Barr virus DNA sequences in human gliomas: Utilizes the Phenol – chloroform – isoamyl alcohol mixture for DNA extraction in medical research, facilitating the study of viral presence in cancer tissues and its implications for oncology and virology (Fonseca et al., 2015).

  • The critical role of DNA extraction for detection of mycobacteria in tissues: This study underscores the effectiveness of the Phenol – chloroform – isoamyl alcohol mixture in the molecular detection of pathogens, serving as a fundamental technique in microbiological diagnostics and infectious disease research (Radomski et al., 2013).

Physical form

mixture of phenol; chloroform; isoamyl alcohol in the ratio of 125:24:1 (v/v/v); saturated with 2 M NaOAc pH 4

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Carc. 2 - Eye Dam. 1 - Muta. 2 - Repr. 2 - Skin Corr. 1B - STOT RE 1 Oral - STOT RE 2

Target Organs

Liver,Kidney, Nervous system,Kidney,Liver,Skin

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dae-Wook Yang et al.
Cell death and differentiation, 28(1), 233-250 (2020-08-02)
The microtubule network is crucial for cell structure and function. Patronin is a conserved protein involved in protecting the minus end of microtubules. Conversely, Klp10A is a kinesin-like microtubule depolymerase. Here we report the role of Drosophila Patronin and Klp10A
Wiesław Bogdanowicz et al.
Proceedings of the National Academy of Sciences of the United States of America, 106(30), 12279-12282 (2009-07-09)
We report the results of mitochondrial and nuclear DNA analyses of skeletal remains exhumed in 2005 at Frombork Cathedral in Poland, that are thought to be those of Nicolaus Copernicus (1473-1543). The analyzed bone remains were found close to the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service