Skip to Content
Merck
All Photos(2)

Key Documents

76159

Supelco

Methyl palmitate

analytical standard

Synonym(s):

n-Hexadecanoic acid methyl ester, Methyl hexadecanoate, Palmitic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)14CO2CH3
CAS Number:
Molecular Weight:
270.45
Beilstein:
1780973
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.5% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.4512 (lit.)

bp

185 °C/10 mmHg (lit.)

mp

32-35 °C (lit.)

density

0.852 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

functional group

ester

shipped in

ambient

storage temp.

room temp

SMILES string

CCCCCCCCCCCCCCCC(=O)OC

InChI

1S/C17H34O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17(18)19-2/h3-16H2,1-2H3

InChI key

FLIACVVOZYBSBS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl palmitate (MeP) is a high melting point component of biodiesel. It is used in a variety of flavors and fragrances and found abundantly in palm oil-related feedstocks.
Methyl palmitate is a fatty acid methyl ester, which shows potential anti-inflammatory and antifibrotic effects by preventing bleomycin-induced lung inflammation and inhibiting the nuclear factor kappa-β pathway(NF-κB).

Application

Methyl palmitate may be used as an analytical reference standard for the quantification of the analyte in biodiesel samples using chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Basic properties of methyl palmitate-diesel blends
Altaie HAM, et al.
Fuel: The Science and Technology of Fuel and Energy, 193(3), 1-6 (2017)
Solvent free synthesis of methyl palmitate over sulfated zirconia solid acid catalyst
Saravanan K, et al.
Fuel: The Science and Technology of Fuel and Energy, 165(3), 298-305 (2016)
Anti-inflammatory and antifibrotic effects of methyl palmitate
El-Demerdash E, et al.
Toxicology and Applied Pharmacology, 254(3), 238-244 (2011)
Characterization of fatty acid methyl esters in biodiesel using high-performance liquid chromatography
Shang NC, et al.
Journal of the Taiwan Institute of Chemical Engineers, 43(3), 354-359 (2012)
Mitigating crystallization of saturated FAMES (fatty acid methyl esters) in biodiesel. 3. The binary phase behavior of 1, 3-dioleoyl-2-palmitoyl glycerol-Methyl palmitate-A multi-length scale structural elucidation of mechanism responsible for inhibiting FAME crystallization
Mohanan A, et al.
Energy, 86(3), 500-513 (2015)

Protocols

HPLC Analysis of Fatty Acid Methyl Esters (FAMES) on SUPELCOSIL™ LC-18

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service