Skip to Content
Merck
All Photos(1)

Key Documents

73120

Supelco

4-Nitrobenzoyl chloride

for HPLC derivatization, LiChropur, ≥99.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4COCl
CAS Number:
Molecular Weight:
185.56
Beilstein:
473192
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.22

grade

for HPLC derivatization

Quality Level

Assay

≥99.0% (GC)

form

crystals

quality

LiChropur

technique(s)

HPLC: suitable

ign. residue

≤0.05% (as SO4)

bp

202-205 °C/105 mmHg (lit.)

mp

71-74 °C (lit.)
71-74 °C

SMILES string

[O-][N+](=O)c1ccc(cc1)C(Cl)=O

InChI

1S/C7H4ClNO3/c8-7(10)5-1-3-6(4-2-5)9(11)12/h1-4H

InChI key

SKDHHIUENRGTHK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Nitrobenzoyl chloride is a derivatization reagent. It reacts with L-glutamic acid undergoing methylation using formaldehyde, synthesizing N-(4-methylaminobenzoyl)glutaminic acid.

Application

4-Nitrobenzoyl chloride was used in separating polyhydric alcohols using HPLC. It was used in snythesizing cellulose benzotates by adding to cellulose/1-allyl-3-methylimidazolium chloride followed by characterization using FT-IF, 1H-NMR and 13C-NMR spectroscopy. It may be used in synthesizing 1-aryloxyacetyl-4-(4-nitrobenzoyl)-thiosemicarbazides under phase transfer catalysis and microwave irradiation with ammonium thiocyanate and aryloxyacetic acid hydrazides.

Other Notes

Derivatization of hydroxy groups for HPLC

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

215.6 °F - closed cup

Flash Point(C)

102 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Separation of some polyhydric alcohols by high-performance liquid chromatography.
Schwarzenbach, Rolf.
Journal of Chromatography A, 140, 304-309 (1977)
F Nachtmann et al.
Journal of chromatography, 122, 293-303 (1976-07-07)
The separation and quantitative determination of digitalis glycosides by high performance liquid chromatography following derivatization with 4-nitrobenzoylchloride (4-NBC1) is described. The compounds of primary interest were the digitalis glycosides and aglycones of the pharmaceutically important A, B and C series
The synthesis of silica-immobilized 1, 2-naphthoquinone thiosemicarbazone with 3-aminopropyltriethoxy silane and 4-nitrobenzoyl chloride.
Audu, Aramani A.
Reactive Polymers, 10, 3-9 (1989)
Journal of Chromatography A, 136, 279-279 (1977)
Tatsuya Higashi et al.
Steroids, 71(7), 609-617 (2006-04-20)
The liquid chromatography-mass spectrometry (LC-MS) methods were developed and validated for the determination of testosterone (T) in the brain and serum of rats and of 5alpha-androstane-3alpha,17beta-diol (ADIOL), a metabolite of T, in the brain of rats. After derivatization of T

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service