Skip to Content
Merck
All Photos(1)

Documents

73108

Supelco

Elemental Impurities according to ICH Q3D oral, Standard 2

TraceCERT®, (in 10% hydrochloric acid), applicable for testing acc. to USP<232>, Ph.Eur. Gen. Chapter 5.20

Synonym(s):

ICH Q3D Elemental Impurity Analysis

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

Agency

according to ICH Q3D
according to Ph. Eur.
according to USP

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

composition

Au, 100 mg/L
Ir, 100 mg/L
Os, 100 mg/L
Pd, 100 mg/L
Pt, 100 mg/L
Rh, 100 mg/L
Ru, 100 mg/L

concentration

(in 10% hydrochloric acid)

technique(s)

ICP: suitable

format

multi-component solution

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Preparation Note

For the preparation of this CRM only materials of highest purity were used.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Met. Corr. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service