Skip to Content
Merck
All Photos(1)

Key Documents

72210

Sigma-Aldrich

Neutral Red

for microscopy (Bact., Bot., Fl., Hist., Vit.), indicator (pH 6.8-8.0)

Synonym(s):

3-Amino-7-dimethylamino-2-methylphenazine hydrochloride, Basic Red 5, Toluylene red

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H17ClN4
CAS Number:
Molecular Weight:
288.78
Colour Index Number:
50040
Beilstein:
3918740
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
MA.02

grade

for microscopy (Bact., Bot., Fl., Hist., Vit.)
indicator (pH 6.8-8.0)

Quality Level

form

powder or crystals

technique(s)

titration: suitable

pH

6.8-8.0, red to yellow

mp

290 °C (dec.) (lit.)

solubility

water: 0.01 g/10 mL

εmax

≥800 at 535-545 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Cl.CN(C)c1ccc2nc3cc(C)c(N)cc3nc2c1

InChI

1S/C15H16N4.ClH/c1-9-6-13-15(8-11(9)16)18-14-7-10(19(2)3)4-5-12(14)17-13;/h4-8H,16H2,1-3H3;1H

InChI key

PGSADBUBUOPOJS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Neutral red is a vital dye widely used as a cellular pH indicator and is selective for shifts in pH. It is highly lipophilic in nature. It favours membrane potential dependent fluorescence that attributes to changes in hydrophobicity.

Application

Neutral red has been used to stain mouse mammary glands.
Useful as an indicator for preparing neutral red paper, and as a biological stain.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

John D Landua et al.
Journal of mammary gland biology and neoplasia, 14(4), 411-415 (2009-11-26)
Whole mount preparations of mouse mammary glands are useful for evaluating overall changes in growth and morphology, and are essential for detecting and evaluating focal or regionally-localized phenotypes that would be difficult to detect or analyze using other techniques. We
Sylvie V M Tesson et al.
Frontiers in microbiology, 9, 2681-2681 (2018-11-30)
Microalgae are common members of the atmospheric microbial assemblages. Diverse airborne microorganisms are known to produce ice nucleation active (INA) compounds, which catalyze cloud and rain formation, and thus alter cloud properties and their own deposition patterns. While the role
Ayesha Ali et al.
Cell reports, 30(5), 1463-1477 (2020-02-06)
The intact vaginal epithelium is essential for women's reproductive health and provides protection against HIV and sexually transmitted infections. How this epithelium maintains itself remains poorly understood. Here, we used single-cell RNA sequencing (RNA-seq) to define the diverse cell populations
Kun Guo et al.
Biosensors & bioelectronics, 47, 184-189 (2013-04-13)
This study introduces a novel and simple method to covalently graft neutral red (NR) onto carbon surfaces based on spontaneous reduction of in situ generated NR diazonium salts. Immobilization of neutral red on carbon surface was achieved by immersing carbon
Cássio do Nascimento et al.
Microscopy research and technique, 76(2), 126-132 (2012-10-31)
The aim of this randomized crossover investigation was to assess the biofilm formation on two titanium and one zirconia substrates in relation to the topography and surface roughness of the materials. Twenty-four discs specimens for each evaluated material (Machined pure

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service