Skip to Content
Merck
All Photos(1)

Key Documents

63560

Millipore

D-Mannitol

ACS reagent, suitable for microbiology, ≥99.0%

Synonym(s):

Mannite

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14O6
CAS Number:
Molecular Weight:
182.17
Beilstein:
1721898
EC Number:
MDL number:
UNSPSC Code:
41106212
PubChem Substance ID:
NACRES:
NA.74

grade

ACS reagent

Quality Level

Assay

≥99.0% (sum of enantiomers, HPLC)
≥99.0%

form

powder

optical activity

[α]20/D +24.0±1° in borax solution acc. to ACS

ign. residue

≤0.01% (as SO4)

loss

≤0.05% loss on drying, 20 °C (HV)

mp

165-169 °C
167-170 °C (lit.)

solubility

H2O: 0.1 g/mL, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

As: ≤0.1 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg
heavy metals: ≤5 ppm (by ICP-OES)

application(s)

microbiology

SMILES string

OC[C@@H](O)[C@@H](O)[C@H](O)[C@H](O)CO

InChI

1S/C6H14O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3-12H,1-2H2/t3-,4-,5-,6-/m1/s1

InChI key

FBPFZTCFMRRESA-KVTDHHQDSA-N

Looking for similar products? Visit Product Comparison Guide

General description

D-Mannitol is a hexose present in plants, bacteria, algae, and fungi.

Application

D-Mannitol has been used as a component of soya flour mannitol (SFM) medium for culturing Streptomyces.

Biochem/physiol Actions

D-Mannitol finds application in the food and pharmaceutical industry. The interaction of mannitol with neutrophils and monocytes is correlated to the upregulation of immune markers. Mannitol acts as a free-radical scavenger and may be beneficial during ischemia-reperfusion injury. It possesses osmotic diuretic properties and may help in managing cerebral edema and high intracranial pressure (ICP).
A sugar alcohol sweet tastant. Used in sweetness inhibition studies.

Other Notes

Important starting material for the synthesis of many chiral building blocks.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Y. Le Merrer et al.
Tetrahedron Letters, 26, 319-319 (1985)
Mannitol: a review of its clinical uses
Shawkat H, et al.
Continuing education in anaesthesia, critical care & pain, 12(2) (12)
Determination of Oxygen Respiration Rates in Wetted Developmentally Arrested Spores of Streptomyces Species
Fischer M, et al.
Bio-protocol, 4(12) (4)
Rapid Nitrate Reduction Assay with Intact Microbial Cells or Spores
Fischer M, et al.
Bio-protocol, 4(12) (4)
S. Hanessian
Total Synthesis of Natural Products: The Chiron Approach (1983)

Articles

Culture media provides a habitat with suitable nutrients, energy sources, and certain environmental conditions for the growth of microorganisms. The components of the culture media range from simple sugars to peptones, salts, antibiotics, and complex indicators.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service