Skip to Content
Merck
All Photos(1)

Key Documents

63175

Sigma-Aldrich

4-Maleimidobutyric acid N-hydroxysuccinimide ester

≥98.0% (HPLC)

Synonym(s):

4-Maleimidobutyric acid N-succinimidyl ester, N-(γ-Maleimidobutyryloxy)succinimide, N-Succinimidyl 4-maleimidobutyrate, GMBS

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H12N2O6
CAS Number:
Molecular Weight:
280.23
Beilstein:
6427689
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥98.0% (HPLC)

reaction suitability

reagent type: linker

mp

123-129 °C

functional group

NHS ester

storage temp.

2-8°C

SMILES string

O=C1N(CCCC(ON2C(CCC2=O)=O)=O)C(C=C1)=O

InChI

1S/C12H12N2O6/c15-8-3-4-9(16)13(8)7-1-2-12(19)20-14-10(17)5-6-11(14)18/h3-4H,1-2,5-7H2

InChI key

PVGATNRYUYNBHO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

A heterobifunctional cross-linking reagent with amine and sulfhydryl reactivity. Typically, coupled initially to molecules containing primary amines by amide bonds buffered at pH 7.5 (6.5-8.5). Second coupling specific for molecules containing free sulfhydryl by thioether linkage buffered at pH 6.8 (6.5-7.0). Useful for preparation of enzyme immunoconjugates and hapten carrier molecule conjugates. Provides a 7-atom linker.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu Zhang et al.
Cancer biology & therapy, 17(11), 1177-1187 (2016-10-22)
Circulating tumor cells (CTCs) have been proposed to be an active source of metastasis or recurrence of hepatocellular carcinoma (HCC). The enumeration and characterization of CTCs has important clinical significance in recurrence prediction and treatment monitoring in HCC patients. We
Jae-Wan Jung et al.
Proteomics, 6(4), 1110-1120 (2006-01-13)
We modified gold arrays with a glutathione (GSH) surface, and investigated high-throughput protein interactions with a spectral surface plasmon resonance (SPR) biosensor. We fabricated the GSH exterior on gold surfaces by successive modification with aminoethanethiol, 4-maleimidobutyric acid N-hydroxysuccinimide ester and
Yan Zhao et al.
Molecular pharmaceutics, 15(7), 2559-2569 (2018-05-17)
Photodynamic therapy, a procedure that uses a photosensitizer to enable light therapy selectively at diseased sites, remains underutilized in oncological clinic. To further improve its cancer selectivity, we developed a polymeric nanosystem by conjugating a photosensitizer IRDye 700DX (IR700) and
Xiubo Zhao et al.
Journal of the Royal Society, Interface, 9(75), 2457-2467 (2012-05-04)
The specific recognition between monoclonal antibody (anti-human prostate-specific antigen, anti-hPSA) and its antigen (human prostate-specific antigen, hPSA) has promising applications in prostate cancer diagnostics and other biosensor applications. However, because of steric constraints associated with interfacial packing and molecular orientations
Kristin Eisenschmidt et al.
Nucleic acids research, 33(22), 7039-7047 (2005-12-17)
Specific cleavage of large DNA molecules at few sites, necessary for the analysis of genomic DNA or for targeting individual genes in complex genomes, requires endonucleases of extremely high specificity. Restriction endonucleases (REase) that recognize DNA sequences of 4-8 bp

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service