Skip to Content
Merck
All Photos(1)

Key Documents

63107

Sigma-Aldrich

Magnesium oxide

tested according to Ph. Eur., heavy

Synonym(s):

Magnesii oxidum ponderosum

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MgO
CAS Number:
Molecular Weight:
40.30
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.21

Agency

USP/NF
tested according to Ph. Eur.

Quality Level

reaction suitability

reagent type: catalyst
core: magnesium

mp

2852 °C (lit.)

SMILES string

O=[Mg]

InChI

1S/Mg.O

InChI key

CPLXHLVBOLITMK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Crystal orientation of epitaxial film deposited on silicon surface.: Research highlights the role of magnesium oxide as a substrate in the deposition of epitaxial films, crucial for the semiconductor industry, offering potential advancements in electronic device fabrication (Kaneko et al., 2024).

  • Advancing in Cesium Retention: Application of Magnesium Phosphate Cement Composites.: This paper discusses the use of magnesium oxide in magnesium phosphate cement composites for cesium retention, providing a novel approach to nuclear waste management and environmental safety (Gharsallah et al., 2024).

  • Investigation of Crystallization Growth Characteristics of Mg(OH)(2) Crystals under Unconstrained Conditions.: Studies the crystallization process of magnesium hydroxide from magnesium oxide, essential for industrial applications such as wastewater treatment and fire retardants (Lv et al., 2024).

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benjamin Michen et al.
Environmental science & technology, 47(3), 1526-1533 (2013-01-05)
Ceramic filters, working on the depth filtration principle, are known to improve drinking water quality by removing human pathogenic microorganisms from contaminated water. However, these microfilters show no sufficient barrier for viruses having diameters down to 20 nm. Recently, it
Charles T Campbell et al.
Journal of the American Chemical Society, 134(43), 18109-18115 (2012-10-05)
Adsorbed molecules are involved in many reactions on solid surface that are of great technological importance. As such, there has been tremendous effort worldwide to learn how to predict reaction rates and equilibrium constants for reactions involving adsorbed molecules. Theoretical
Maryam Sayyah et al.
ChemSusChem, 6(1), 193-198 (2012-11-08)
The reversible cycling of CaO adsorbents to CaCO(3) for high-temperature CO(2) capture is substantially improved by mechanical treatment. The mechanical milling intensity and conditions of grinding (e.g., wet vs. dry, planetary vs. vibratory milling) were determined to be the main
Magdalena Maciejewska et al.
Journal of chromatography. A, 1257, 141-148 (2012-08-22)
Inverse gas chromatography (IGC) was applied at infinite dilution to evaluate the surface properties of calcium and magnesium oxide nanoparticles and the effect of surface grafted unsaturated carboxylic acid on the nanopowder donor-acceptor characteristics. The dispersive components (γ(s)(D)) of the
Ali Jebali et al.
Toxicology in vitro : an international journal published in association with BIBRA, 27(6), 1896-1904 (2013-06-29)
Cutaneous leishmaniasis (CL) is endemic in the tropical and subtropical countries. Antileishmanial drugs that are traditionally used for treatment of CL are mainly toxic, ineffective for some parasite isolates, and mostly expensive. Previous studies showed that some metal and metal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service