Skip to Content
Merck
All Photos(1)

Key Documents

60063

Sigma-Aldrich

Potassium antimonyl tartrate trihydrate

purum p.a., 99.0-103% (RT)

Synonym(s):

Antimony potassium tartrate trihydrate, Tartar emetic

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H4K2O12Sb2 · 3H2O
CAS Number:
Molecular Weight:
667.87
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

grade

purum p.a.

Quality Level

Assay

99.0-103% (RT)

loss

≤2.7% loss on drying

mp

≥300 °C (lit.)

anion traces

chloride (Cl-): ≤100 mg/kg
sulfate (SO42-): ≤500 mg/kg

cation traces

Ca: ≤50 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
Na: ≤500 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

O.O.O.[K+].[K+].O=C1O[Sb-]23OC1C4O[Sb-]5(OC(C(O2)C(=O)O3)C(=O)O5)OC4=O

InChI

1S/2C4H4O6.2K.3H2O.2Sb/c2*5-1(3(7)8)2(6)4(9)10;;;;;;;/h2*1-2H,(H,7,8)(H,9,10);;;3*1H2;;/q2*-2;2*+1;;;;2*+3/p-4

InChI key

WBTCZEPSIIFINA-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

General description

Potassium antimonyl tartrate trihydrate is a trivalent antimony compound. It has been reported to be a leishmanicidal compound and exhibits more toxic action than pentavalent antimony (Pentostam) against Leishmania species.[1][2] Three-dimensional X-ray and white radiation neutron diffraction methods have been reported to investigate the crystal structure of its optically active form (dipotassium di-μ-d-tartrato (4)-bis(antimonate(III)) trihydrate). Unit cell dimensions reported were: a = 11.192(2), b = 11.696(3), and c = 25.932(5)Å.[3]

Application

Potassium antimonyl tartrate trihydrate may be used as an Sb(III)-containing drug to investigate its ability to induce cell death associated with DNA fragmentation in axenic amastigotes of Leishmania infantum.[4]

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Sereno et al.
Antimicrobial agents and chemotherapy, 45(7), 2064-2069 (2001-06-16)
The basic treatment of leishmaniasis consists in the administration of pentavalent antimonials. The mechanisms that contribute to pentavalent antimonial toxicity against the intracellular stage of the parasite (i.e., amastigote) are still unknown. In this study, the combined use of several
D Sereno et al.
Antimicrobial agents and chemotherapy, 41(5), 972-976 (1997-05-01)
Using a 3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide microassay, previously described as a means of quantifying Leishmania amazonensis in vitro at the amastigote stage (D. Sereno and J. L. Lemesre, Parisitol. Res., in press), we have compared the activities of seven drugs, including those
Daniel B Liarte et al.
Parasitology research, 107(1), 205-212 (2010-04-08)
In the present study, we selected in vitro populations of Leishmania Viannia guyanensis, L.V. braziliensis, L. Leishmania amazonensis and L.L. infantum chagasi that were resistant to potassium antimony tartrate (SbIII). The resistance index of these populations varied from 4- to
Zahra Faghiri et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 23(8), 2780-2789 (2009-04-15)
Schistosomes are parasitic platyhelminths that constitute an important public health problem globally. Infection is characterized by the presence of adult worms within the vasculature of their hosts, where they can reside for many years. The worms are covered by an
Avijit Sarkar et al.
Cytometry. Part A : the journal of the International Society for Analytical Cytology, 79(1), 35-45 (2010-12-25)
Nitric oxide (NO) has been demonstrated to be a principal effector molecule responsible for mediating intracellular killing of Leishmania parasites, the causative organism of leishmaniasis. As measurement of intracellular NO remains a challenge to biologists, we have developed a flow

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service