Skip to Content
Merck
All Photos(1)

Key Documents

50194

Supelco

Sulisobenzone

analytical standard

Synonym(s):

5-Benzoyl-4-hydroxy-2-methoxybenzenesulfonic acid, HMBS, Sulisobenzone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H12O6S
CAS Number:
Molecular Weight:
308.31
Beilstein:
2889165
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.5% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

COc1cc(O)c(cc1S(O)(=O)=O)C(=O)c2ccccc2

InChI

1S/C14H12O6S/c1-20-12-8-11(15)10(7-13(12)21(17,18)19)14(16)9-5-3-2-4-6-9/h2-8,15H,1H3,(H,17,18,19)

InChI key

CXVGEDCSTKKODG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sulisobenzone is a broad spectrum organic UV filter in sunscreen formulations, known to absorb deleterious UV light.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Sulisobenzone may be used as an analytical reagent for the determination of the analyte in sunscreen formulations by high-performance liquid chromatography with UV spectrophotometric detection.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J S Azevedo et al.
Farmaco (Societa chimica italiana : 1989), 54(9), 573-578 (1999-11-11)
UVA/UVB protective agents are important constituents in many cosmetics and sunscreen preparations. Analytical methodologies which enable the determination of combined substances with different spectrophotometric characteristics are therefore useful and cost-effective. To this end, the following substances in a sunscreen gel
Z León et al.
Analytical and bioanalytical chemistry, 391(3), 859-866 (2008-03-28)
An analytical method based on ion-interaction chromatography with UV detection for simultaneous in-vitro estimation of the percutaneous absorption of the most used water-soluble UV filters in sunscreen cosmetics is proposed. These UV filters were phenylbenzimidazole sulfonic acid, disodium phenyl dibenzimidazole
Anna Bianchi et al.
Journal of pharmaceutical and biomedical analysis, 56(4), 692-697 (2011-08-03)
The aim of the study was to examine the photostability of the major catechin of green tea, (-)-epigallocatechin-3-gallate (EGCG), which possesses important antioxidant and skin photoprotective properties. In order to simulate realistic conditions of use of topical preparations, the photolysis
Satomi Onoue et al.
Journal of pharmaceutical and biomedical analysis, 47(4-5), 967-972 (2008-05-06)
We have previously demonstrated that the phototoxic potential of chemicals could be partly predicted by the determination of reactive oxygen species (ROS) from photo-irradiated compounds. In this study, ROS assay strategy was applied to 39 marketed drugs and 210 drug
N Negreira et al.
Analytica chimica acta, 743, 101-110 (2012-08-14)
The stability of the UV filter benzophenone-4 (BP-4) in free chlorine-containing water was investigated, for the first time, by liquid chromatography quadrupole time-of-flight mass spectrometry (LC-QqTOF-MS). High mass accuracy and resolution capabilities of this hybrid mass spectrometer were used for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service