Skip to Content
Merck
All Photos(1)

Key Documents

49825

Millipore

DMACA Reagent

suitable for microbiology

Synonym(s):

4-(Dimethylamino)-cinnamaldehyde solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H13NO
CAS Number:
Molecular Weight:
175.23
Beilstein:
2208002
MDL number:
UNSPSC Code:
41171621
PubChem Substance ID:
NACRES:
NA.85

technique(s)

microbe id | specific enzyme detection: suitable

Quality Level

application(s)

agriculture
environmental
food and beverages

microbiology

suitability

Escherichia coli
Streptococcus spp.

SMILES string

[H]C(=O)\C=C\c1ccc(cc1)N(C)C

InChI

1S/C11H13NO/c1-12(2)11-7-5-10(6-8-11)4-3-9-13/h3-9H,1-2H3/b4-3+

InChI key

RUKJCCIJLIMGEP-ONEGZZNKSA-N

General description

DMACA is basically a selective staining reagent. It helps identify the flavanols by staining them blue in color.

Application

Used to determine the ability of an organism to split indole from the tryptophan molecule.

Components

Composition per 100 ml:
Dimethylaminocinnamaldehye 1 g
Hydrochloric acid (concentrated) 1.0 ml
distilled water 99.0 ml

Other Notes

Reagent for the rapid identification of Streptococci.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stephen J Robinson et al.
BMC plant biology, 9, 101-101 (2009-08-04)
Functional genomics tools provide researchers with the ability to apply high-throughput techniques to determine the function and interaction of a diverse range of genes. Mutagenized plant populations are one such resource that facilitate gene characterisation. They allow complex physiological responses
Normi Izati Mat Nawi et al.
Polymers, 11(5) (2019-05-16)
Membrane distillation (MD) is an attractive technology for desalination, mainly because its performance that is almost independent of feed solute concentration as opposed to the reverse osmosis process. However, its widespread application is still limited by the low water flux
Sihui Ma et al.
Journal of food science, 84(8), 2147-2158 (2019-07-18)
Multiple analytical methods are used for quantification of total polyphenols and total flavanols in fruit juices and beverages. Four methods were evaluated in this study: Folin-Ciocalteu (F-C), Lowenthal permanganate (L-P), 4-dimethylaminocinnamaldehyde (DMAC), and the bovine serum albumin (BSA) precipitation method.
J.F. MacFaddin
Biochemical Tests for Identification of Medical Bacteria (2000)
Camille Melissa Johnston et al.
BMC genomics, 19(1), 600-600 (2018-08-11)
Direct molecular cloning of full-length cDNAs derived from viral RNA is an approach to identify the individual viral genomes within a virus population. This enables characterization of distinct viral haplotypes present during infection. In this study, we recover individual genomes

Articles

There are many other methods of detection to indicate the presence of E. coli. Review common tests and biochemical reactions for this contaminant.

There are many other methods of detection to indicate the presence of E. coli. Review common tests and biochemical reactions for this contaminant.

There are many other methods of detection to indicate the presence of E. coli. Review common tests and biochemical reactions for this contaminant.

There are many other methods of detection to indicate the presence of E. coli. Review common tests and biochemical reactions for this contaminant.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service